Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 9245459-9245684 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
H6PD
Entrez Gene ID
9563
Gene Name
hexose-6-phosphate dehydrogenase/glucose 1-dehydrogenase
Gene Aliases
CORTRD1, G6PDH, GDH
UniGene
Hs.463511
Gene Chromosome Location
Chr.1: 9234767-9271337 on Build GRCh38
Transcripts
NM_001282587.1 NM_004285.3 XM_005263540.4 XM_006711052.3 XM_017002865.1 XM_017002866.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3493369 c.609C>T p.D203D Chr.1: 9245543-9245543 on Build GRCh38
1665922 c.627+1G>C p.? Chr.1: 9245562-9245562 on Build GRCh38
4010629 c.627+2T>C p.? Chr.1: 9245563-9245563 on Build GRCh38
4552652 c.565G>A p.E189K Chr.1: 9245499-9245499 on Build GRCh38
913321 c.626A>G p.Q209R Chr.1: 9245560-9245560 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200223725 Chr.1: 9245533-9245541 on Build GRCh38
rs201667735 Chr.1: 9245520-9245520 on Build GRCh38
rs572686829 Chr.1: 9245634-9245634 on Build GRCh38
rs372000451 Chr.1: 9245599-9245599 on Build GRCh38
rs777494312 Chr.1: 9245546-9245549 on Build GRCh38
rs752784177 Chr.1: 9245592-9245592 on Build GRCh38
rs538609204 Chr.1: 9245653-9245653 on Build GRCh38
rs376601311 Chr.1: 9245605-9245605 on Build GRCh38
rs375492775 Chr.1: 9245641-9245641 on Build GRCh38
rs148507306 Chr.1: 9245536-9245536 on Build GRCh38
rs200086283 Chr.1: 9245490-9245490 on Build GRCh38
rs764642458 Chr.1: 9245600-9245600 on Build GRCh38
rs765256828 Chr.1: 9245583-9245583 on Build GRCh38
rs142705762 Chr.1: 9245607-9245607 on Build GRCh38
rs999834759 Chr.1: 9245575-9245575 on Build GRCh38
rs774134939 Chr.1: 9245518-9245518 on Build GRCh38
rs772023941 Chr.1: 9245616-9245616 on Build GRCh38
rs1016412474 Chr.1: 9245655-9245655 on Build GRCh38
rs369633008 Chr.1: 9245631-9245631 on Build GRCh38
rs910028601 Chr.1: 9245651-9245651 on Build GRCh38
rs950404089 Chr.1: 9245635-9245635 on Build GRCh38
rs771839732 Chr.1: 9245535-9245535 on Build GRCh38
rs558537168 Chr.1: 9245598-9245598 on Build GRCh38
rs758114149 Chr.1: 9245518-9245520 on Build GRCh38
rs757758611 Chr.1: 9245606-9245606 on Build GRCh38
rs749203003 Chr.1: 9245513-9245513 on Build GRCh38
rs201196113 Chr.1: 9245580-9245580 on Build GRCh38
rs759114595 Chr.1: 9245630-9245630 on Build GRCh38
rs768539842 Chr.1: 9245514-9245514 on Build GRCh38
rs149366674 Chr.1: 9245504-9245504 on Build GRCh38

View Full Product Details