Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 98716157-98716426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf177
Entrez Gene ID
283598
Gene Name
chromosome 14 open reading frame 177
Gene Aliases
-
UniGene
Hs.448754
Gene Chromosome Location
Chr.14: 98711613-98717766 on Build GRCh38
Transcripts
NM_182560.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
369898 c.116G>T p.G39V Chr.14: 98716307-98716307 on Build GRCh38
5848840 c.191G>T p.R64M Chr.14: 98716382-98716382 on Build GRCh38
3499619 c.117G>A p.G39G Chr.14: 98716308-98716308 on Build GRCh38
6141685 c.40A>G p.R14G Chr.14: 98716231-98716231 on Build GRCh38
6076763 c.172C>A p.L58M Chr.14: 98716363-98716363 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs930417275 Chr.14: 98716288-98716288 on Build GRCh38
rs373583218 Chr.14: 98716193-98716201 on Build GRCh38
rs746548927 Chr.14: 98716389-98716389 on Build GRCh38
rs865870167 Chr.14: 98716207-98716207 on Build GRCh38
rs754834530 Chr.14: 98716369-98716369 on Build GRCh38
rs747793829 Chr.14: 98716314-98716314 on Build GRCh38
rs751661133 Chr.14: 98716249-98716249 on Build GRCh38
rs771714983 Chr.14: 98716378-98716378 on Build GRCh38
rs1019281452 Chr.14: 98716253-98716253 on Build GRCh38
rs772790411 Chr.14: 98716322-98716322 on Build GRCh38
rs201129643 Chr.14: 98716186-98716186 on Build GRCh38
rs754064293 Chr.14: 98716197-98716197 on Build GRCh38
rs17097718 Chr.14: 98716198-98716198 on Build GRCh38
rs747879883 Chr.14: 98716377-98716377 on Build GRCh38
rs1020823666 Chr.14: 98716206-98716206 on Build GRCh38
rs769793259 Chr.14: 98716189-98716190 on Build GRCh38
rs751695178 Chr.14: 98716356-98716356 on Build GRCh38
rs763286311 Chr.14: 98716340-98716340 on Build GRCh38
rs761230662 Chr.14: 98716270-98716270 on Build GRCh38
rs748267539 Chr.14: 98716199-98716199 on Build GRCh38
rs529559865 Chr.14: 98716315-98716315 on Build GRCh38
rs143017999 Chr.14: 98716307-98716307 on Build GRCh38
rs370871652 Chr.14: 98716258-98716258 on Build GRCh38
rs766761695 Chr.14: 98716196-98716196 on Build GRCh38
rs777231040 Chr.14: 98716397-98716397 on Build GRCh38
rs8009159 Chr.14: 98716222-98716222 on Build GRCh38
rs755181814 Chr.14: 98716391-98716392 on Build GRCh38
rs771497184 Chr.14: 98716330-98716330 on Build GRCh38
rs373786215 Chr.14: 98716327-98716327 on Build GRCh38
rs559757171 Chr.14: 98716373-98716373 on Build GRCh38
rs906789253 Chr.14: 98716316-98716316 on Build GRCh38
rs763599935 Chr.14: 98716181-98716181 on Build GRCh38
rs751741080 Chr.14: 98716391-98716394 on Build GRCh38
rs534411159 Chr.14: 98716235-98716235 on Build GRCh38
rs200211837 Chr.14: 98716391-98716391 on Build GRCh38
rs371078399 Chr.14: 98716361-98716361 on Build GRCh38
rs762657048 Chr.14: 98716179-98716179 on Build GRCh38
rs773773781 Chr.14: 98716269-98716269 on Build GRCh38
rs373945676 Chr.14: 98716268-98716268 on Build GRCh38
rs145620507 Chr.14: 98716272-98716272 on Build GRCh38
rs758498998 Chr.14: 98716211-98716211 on Build GRCh38
rs574306596 Chr.14: 98716392-98716392 on Build GRCh38
rs202177322 Chr.14: 98716238-98716238 on Build GRCh38
rs764348134 Chr.14: 98716346-98716346 on Build GRCh38
rs756084042 Chr.14: 98716298-98716298 on Build GRCh38
rs768387385 Chr.14: 98716257-98716257 on Build GRCh38
rs539855636 Chr.14: 98716185-98716185 on Build GRCh38
rs893793862 Chr.14: 98716344-98716344 on Build GRCh38
rs765575373 Chr.14: 98716284-98716284 on Build GRCh38
rs151268972 Chr.14: 98716209-98716209 on Build GRCh38
rs533269718 Chr.14: 98716383-98716383 on Build GRCh38
rs777990786 Chr.14: 98716290-98716290 on Build GRCh38
rs759990024 Chr.14: 98716337-98716337 on Build GRCh38
rs562246145 Chr.14: 98716313-98716313 on Build GRCh38
rs768419055 Chr.14: 98716311-98716311 on Build GRCh38
rs368720378 Chr.14: 98716333-98716333 on Build GRCh38
rs573543125 Chr.14: 98716232-98716232 on Build GRCh38
rs751540018 Chr.14: 98716295-98716295 on Build GRCh38
rs759775875 Chr.14: 98716280-98716280 on Build GRCh38
rs200985291 Chr.14: 98716279-98716279 on Build GRCh38
rs761362803 Chr.14: 98716259-98716259 on Build GRCh38
rs1955907 Chr.14: 98716339-98716339 on Build GRCh38
rs145157665 Chr.14: 98716368-98716368 on Build GRCh38
rs139827156 Chr.14: 98716190-98716198 on Build GRCh38
rs938270789 Chr.14: 98716336-98716336 on Build GRCh38
rs780090156 Chr.14: 98716367-98716367 on Build GRCh38
rs4905757 Chr.14: 98716289-98716289 on Build GRCh38
rs577677223 Chr.14: 98716261-98716261 on Build GRCh38
rs370033261 Chr.14: 98716306-98716306 on Build GRCh38
rs756474357 Chr.14: 98716187-98716187 on Build GRCh38

View Full Product Details