Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67281655-67281910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRK2
Entrez Gene ID
156
Gene Name
G protein-coupled receptor kinase 2
Gene Aliases
ADRBK1, BARK1, BETA-ARK1
UniGene
Hs.83636
Gene Chromosome Location
Chr.11: 67266420-67286558 on Build GRCh38
Transcripts
NM_001619.3 XM_011544773.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6070268 c.826_826+1GG>CA p.? Chr.11: 67281728-67281729 on Build GRCh38
5373364 c.827G>A p.G276D Chr.11: 67281822-67281822 on Build GRCh38
5589571 c.849C>T p.L283L Chr.11: 67281844-67281844 on Build GRCh38
6000057 c.897C>T p.A299A Chr.11: 67281892-67281892 on Build GRCh38
1317270 c.827-2A>G p.? Chr.11: 67281820-67281820 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201089866 Chr.11: 67281727-67281727 on Build GRCh38
rs374493600 Chr.11: 67281758-67281758 on Build GRCh38
rs753729641 Chr.11: 67281746-67281746 on Build GRCh38
rs543467610 Chr.11: 67281690-67281690 on Build GRCh38
rs756737110 Chr.11: 67281884-67281884 on Build GRCh38
rs770069873 Chr.11: 67281738-67281738 on Build GRCh38
rs374978008 Chr.11: 67281879-67281879 on Build GRCh38
rs746366121 Chr.11: 67281761-67281761 on Build GRCh38
rs367914738 Chr.11: 67281889-67281889 on Build GRCh38
rs142106623 Chr.11: 67281878-67281878 on Build GRCh38
rs138298425 Chr.11: 67281742-67281742 on Build GRCh38
rs947074070 Chr.11: 67281700-67281700 on Build GRCh38
rs148898193 Chr.11: 67281679-67281679 on Build GRCh38
rs781544095 Chr.11: 67281697-67281697 on Build GRCh38
rs759410424 Chr.11: 67281807-67281807 on Build GRCh38
rs765043700 Chr.11: 67281751-67281751 on Build GRCh38
rs780591835 Chr.11: 67281765-67281765 on Build GRCh38
rs1022669933 Chr.11: 67281730-67281730 on Build GRCh38
rs780679941 Chr.11: 67281885-67281885 on Build GRCh38
rs545272790 Chr.11: 67281774-67281774 on Build GRCh38
rs756649187 Chr.11: 67281764-67281764 on Build GRCh38
rs751186389 Chr.11: 67281875-67281875 on Build GRCh38
rs962248596 Chr.11: 67281743-67281743 on Build GRCh38
rs377123753 Chr.11: 67281682-67281682 on Build GRCh38
rs749651599 Chr.11: 67281768-67281768 on Build GRCh38
rs373462879 Chr.11: 67281741-67281741 on Build GRCh38
rs885833 Chr.11: 67281745-67281745 on Build GRCh38
rs762730044 Chr.11: 67281868-67281868 on Build GRCh38
rs370779125 Chr.11: 67281787-67281787 on Build GRCh38
rs1041374864 Chr.11: 67281703-67281703 on Build GRCh38
rs202054233 Chr.11: 67281872-67281872 on Build GRCh38
rs765789108 Chr.11: 67281795-67281797 on Build GRCh38
rs576089239 Chr.11: 67281688-67281688 on Build GRCh38
rs150930474 Chr.11: 67281859-67281859 on Build GRCh38
rs775930888 Chr.11: 67281739-67281739 on Build GRCh38
rs755384357 Chr.11: 67281888-67281888 on Build GRCh38
rs752437203 Chr.11: 67281755-67281755 on Build GRCh38
rs142524893 Chr.11: 67281856-67281856 on Build GRCh38
rs774712563 Chr.11: 67281776-67281776 on Build GRCh38
rs777286637 Chr.11: 67281686-67281686 on Build GRCh38
rs369546263 Chr.11: 67281740-67281740 on Build GRCh38
rs749739399 Chr.11: 67281886-67281886 on Build GRCh38
rs772046568 Chr.11: 67281788-67281788 on Build GRCh38
rs3730143 Chr.11: 67281771-67281771 on Build GRCh38
rs773299198 Chr.11: 67281797-67281797 on Build GRCh38
rs372402791 Chr.11: 67281757-67281757 on Build GRCh38
rs758003046 Chr.11: 67281685-67281685 on Build GRCh38
rs765135845 Chr.11: 67281853-67281853 on Build GRCh38
rs7350469 Chr.11: 67281830-67281830 on Build GRCh38
rs753710294 Chr.11: 67281680-67281680 on Build GRCh38
rs756869266 Chr.11: 67281876-67281876 on Build GRCh38
rs746303623 Chr.11: 67281737-67281737 on Build GRCh38
rs760039738 Chr.11: 67281738-67281754 on Build GRCh38
rs759448861 Chr.11: 67281747-67281747 on Build GRCh38
rs771507227 Chr.11: 67281691-67281691 on Build GRCh38
rs200171288 Chr.11: 67281759-67281759 on Build GRCh38

View Full Product Details