Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45771762-45771987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMPK
Entrez Gene ID
1760
Gene Name
dystrophia myotonica protein kinase
Gene Aliases
DM, DM1, DM1PK, DMK, MDPK, MT-PK
UniGene
Hs.631596
Gene Chromosome Location
Chr.19: 45769709-45782557 on Build GRCh38
Transcripts
NM_001081560.2 NM_001081562.2 NM_001081563.2 NM_001288764.1 NM_001288765.1 NM_001288766.1 NM_004409.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6856143 c.1406C>T p.A469V Chr.19: 45771897-45771897 on Build GRCh38
6429867 c.1379A>G p.E460G Chr.19: 45771924-45771924 on Build GRCh38
6645650 c.1407G>A p.R469R Chr.19: 45771866-45771866 on Build GRCh38
4650504 c.1400C>T p.T467M Chr.19: 45771873-45771873 on Build GRCh38
6038228 c.1384G>A p.A462T Chr.19: 45771919-45771919 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541123797 Chr.19: 45771939-45771939 on Build GRCh38
rs374095905 Chr.19: 45771779-45771779 on Build GRCh38
rs187454872 Chr.19: 45771915-45771915 on Build GRCh38
rs774925987 Chr.19: 45771941-45771941 on Build GRCh38
rs749309049 Chr.19: 45771802-45771802 on Build GRCh38
rs147014815 Chr.19: 45771881-45771881 on Build GRCh38
rs866132084 Chr.19: 45771806-45771806 on Build GRCh38
rs751708764 Chr.19: 45771785-45771785 on Build GRCh38
rs498916 Chr.19: 45771935-45771935 on Build GRCh38
rs755023780 Chr.19: 45771894-45771894 on Build GRCh38
rs78771765 Chr.19: 45771796-45771796 on Build GRCh38
rs894484802 Chr.19: 45771920-45771920 on Build GRCh38
rs930728803 Chr.19: 45771898-45771898 on Build GRCh38
rs138229648 Chr.19: 45771906-45771906 on Build GRCh38
rs763358330 Chr.19: 45771873-45771873 on Build GRCh38
rs554723850 Chr.19: 45771813-45771813 on Build GRCh38
rs760107585 Chr.19: 45771838-45771838 on Build GRCh38
rs745695704 Chr.19: 45771913-45771913 on Build GRCh38
rs752693597 Chr.19: 45771899-45771899 on Build GRCh38
rs372615381 Chr.19: 45771948-45771948 on Build GRCh38
rs745642787 Chr.19: 45771814-45771814 on Build GRCh38
rs778777182 Chr.19: 45771910-45771910 on Build GRCh38
rs771735401 Chr.19: 45771827-45771827 on Build GRCh38
rs770443048 Chr.19: 45771952-45771952 on Build GRCh38
rs987706871 Chr.19: 45771820-45771820 on Build GRCh38
rs141405839 Chr.19: 45771869-45771869 on Build GRCh38
rs370875536 Chr.19: 45771940-45771940 on Build GRCh38
rs147824333 Chr.19: 45771830-45771830 on Build GRCh38
rs112681257 Chr.19: 45771895-45771895 on Build GRCh38
rs762450145 Chr.19: 45771867-45771867 on Build GRCh38
rs188834458 Chr.19: 45771850-45771850 on Build GRCh38
rs374509696 Chr.19: 45771956-45771956 on Build GRCh38
rs765628468 Chr.19: 45771868-45771868 on Build GRCh38
rs768189290 Chr.19: 45771938-45771938 on Build GRCh38
rs775989938 Chr.19: 45771864-45771864 on Build GRCh38
rs200807652 Chr.19: 45771792-45771792 on Build GRCh38
rs761415826 Chr.19: 45771897-45771897 on Build GRCh38
rs779695594 Chr.19: 45771923-45771923 on Build GRCh38
rs747588213 Chr.19: 45771950-45771950 on Build GRCh38
rs866838143 Chr.19: 45771808-45771808 on Build GRCh38

View Full Product Details