Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3190740-3190971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDRGK1
Entrez Gene ID
65992
Gene Name
DDRGK domain containing 1
Gene Aliases
C20orf116, UFBP1, dJ1187M17.3
UniGene
Hs.471975
Gene Chromosome Location
Chr.20: 3190350-3204697 on Build GRCh38
Transcripts
NM_023935.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7157598 c.857G>A p.R286Q Chr.20: 3190741-3190741 on Build GRCh38
4510725 c.846C>T p.F282F Chr.20: 3190752-3190752 on Build GRCh38
6421764 c.835G>A p.V279M Chr.20: 3190763-3190763 on Build GRCh38
5629394 c.785T>C p.I262T Chr.20: 3190813-3190813 on Build GRCh38
2757300 c.791A>G p.D264G Chr.20: 3190807-3190807 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754900499 Chr.20: 3190766-3190766 on Build GRCh38
rs1055113251 Chr.20: 3190900-3190900 on Build GRCh38
rs767124243 Chr.20: 3190841-3190841 on Build GRCh38
rs1052913469 Chr.20: 3190780-3190780 on Build GRCh38
rs781609753 Chr.20: 3190805-3190805 on Build GRCh38
rs6139028 Chr.20: 3190849-3190849 on Build GRCh38
rs904437548 Chr.20: 3190930-3190930 on Build GRCh38
rs185570984 Chr.20: 3190836-3190836 on Build GRCh38
rs939446921 Chr.20: 3190897-3190897 on Build GRCh38
rs1035810320 Chr.20: 3190940-3190940 on Build GRCh38
rs771277106 Chr.20: 3190787-3190787 on Build GRCh38
rs376415173 Chr.20: 3190808-3190808 on Build GRCh38
rs3171339 Chr.20: 3190791-3190791 on Build GRCh38
rs895177331 Chr.20: 3190918-3190918 on Build GRCh38
rs953670088 Chr.20: 3190811-3190811 on Build GRCh38
rs34321897 Chr.20: 3190783-3190784 on Build GRCh38
rs750123184 Chr.20: 3190851-3190851 on Build GRCh38
rs6139027 Chr.20: 3190835-3190835 on Build GRCh38
rs374875243 Chr.20: 3190929-3190929 on Build GRCh38
rs760339077 Chr.20: 3190858-3190858 on Build GRCh38
rs984394162 Chr.20: 3190821-3190821 on Build GRCh38
rs946706289 Chr.20: 3190920-3190920 on Build GRCh38
rs928122275 Chr.20: 3190890-3190890 on Build GRCh38
rs779943089 Chr.20: 3190762-3190762 on Build GRCh38
rs755941311 Chr.20: 3190760-3190760 on Build GRCh38
rs142248347 Chr.20: 3190764-3190764 on Build GRCh38
rs116920518 Chr.20: 3190942-3190942 on Build GRCh38
rs770238423 Chr.20: 3190809-3190809 on Build GRCh38
rs763451293 Chr.20: 3190813-3190813 on Build GRCh38
rs920944740 Chr.20: 3190799-3190799 on Build GRCh38
rs777838925 Chr.20: 3190767-3190767 on Build GRCh38
rs769057983 Chr.20: 3190826-3190826 on Build GRCh38
rs747069299 Chr.20: 3190777-3190777 on Build GRCh38
rs998744202 Chr.20: 3190946-3190946 on Build GRCh38
rs910119871 Chr.20: 3190855-3190855 on Build GRCh38
rs375033804 Chr.20: 3190861-3190861 on Build GRCh38
rs766155917 Chr.20: 3190860-3190860 on Build GRCh38
rs995195689 Chr.20: 3190875-3190875 on Build GRCh38

View Full Product Details