Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 23896946-23897201 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QTNF9B
Entrez Gene ID
387911
Gene Name
C1q and tumor necrosis factor related protein 9B
Gene Aliases
CTRP9B
UniGene
Hs.740810
Gene Chromosome Location
Chr.13: 23891099-23898541 on Build GRCh38
Transcripts
NM_001007537.2 NR_104426.1 XM_011535070.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6423915 c.16C>T p.L6F Chr.13: 23896971-23896971 on Build GRCh38
5362131 c.5G>T p.R2M Chr.13: 23896982-23896982 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs9510930 Chr.13: 23897093-23897093 on Build GRCh38
rs775453546 Chr.13: 23896984-23896984 on Build GRCh38
rs767257524 Chr.13: 23897165-23897165 on Build GRCh38
rs535719611 Chr.13: 23897072-23897072 on Build GRCh38
rs112955587 Chr.13: 23897003-23897004 on Build GRCh38
rs767209031 Chr.13: 23897012-23897012 on Build GRCh38
rs750847065 Chr.13: 23897021-23897021 on Build GRCh38
rs752457756 Chr.13: 23897170-23897170 on Build GRCh38
rs981605769 Chr.13: 23897121-23897121 on Build GRCh38
rs201194245 Chr.13: 23896971-23896971 on Build GRCh38
rs191606407 Chr.13: 23897173-23897173 on Build GRCh38
rs1012994001 Chr.13: 23897100-23897100 on Build GRCh38
rs186828471 Chr.13: 23897058-23897058 on Build GRCh38
rs111568876 Chr.13: 23897071-23897071 on Build GRCh38
rs558014120 Chr.13: 23897124-23897124 on Build GRCh38
rs780474046 Chr.13: 23897031-23897031 on Build GRCh38
rs751311504 Chr.13: 23897007-23897007 on Build GRCh38
rs111654463 Chr.13: 23897076-23897076 on Build GRCh38
rs548924153 Chr.13: 23897064-23897064 on Build GRCh38
rs370628710 Chr.13: 23897111-23897111 on Build GRCh38
rs1032972408 Chr.13: 23897078-23897078 on Build GRCh38
rs139097033 Chr.13: 23896983-23896983 on Build GRCh38
rs971991398 Chr.13: 23897119-23897119 on Build GRCh38
rs764753254 Chr.13: 23897060-23897060 on Build GRCh38
rs762422936 Chr.13: 23896985-23896985 on Build GRCh38
rs961614233 Chr.13: 23897160-23897160 on Build GRCh38
rs756451157 Chr.13: 23897025-23897025 on Build GRCh38
rs181995253 Chr.13: 23897008-23897008 on Build GRCh38

View Full Product Details