Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1452288-1452561 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATAD3C
Entrez Gene ID
219293
Gene Name
ATPase family, AAA domain containing 3C
Gene Aliases
-
UniGene
Hs.724767
Gene Chromosome Location
Chr.1: 1449689-1470158 on Build GRCh38
Transcripts
NM_001039211.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
236634 c.203G>A p.R68Q Chr.1: 1452415-1452415 on Build GRCh38
3472798 c.189C>T p.A63A Chr.1: 1452401-1452401 on Build GRCh38
6606424 c.157G>A p.A53T Chr.1: 1452369-1452369 on Build GRCh38
3862508 c.184C>T p.R62C Chr.1: 1452396-1452396 on Build GRCh38
6452023 c.222G>A p.T74T Chr.1: 1452434-1452434 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555532088 Chr.1: 1452473-1452473 on Build GRCh38
rs9793066 Chr.1: 1452346-1452346 on Build GRCh38
rs745620239 Chr.1: 1452392-1452392 on Build GRCh38
rs9793256 Chr.1: 1452384-1452384 on Build GRCh38
rs769925964 Chr.1: 1452358-1452359 on Build GRCh38
rs956743914 Chr.1: 1452407-1452407 on Build GRCh38
rs755836496 Chr.1: 1452444-1452444 on Build GRCh38
rs761074299 Chr.1: 1452416-1452416 on Build GRCh38
rs113110734 Chr.1: 1452367-1452367 on Build GRCh38
rs980403508 Chr.1: 1452453-1452453 on Build GRCh38
rs745673167 Chr.1: 1452442-1452442 on Build GRCh38
rs768274901 Chr.1: 1452406-1452406 on Build GRCh38
rs773725479 Chr.1: 1452409-1452409 on Build GRCh38
rs764809800 Chr.1: 1452519-1452519 on Build GRCh38
rs748797994 Chr.1: 1452405-1452405 on Build GRCh38
rs780385481 Chr.1: 1452341-1452341 on Build GRCh38
rs888100049 Chr.1: 1452368-1452368 on Build GRCh38
rs766918342 Chr.1: 1452472-1452472 on Build GRCh38
rs535410015 Chr.1: 1452427-1452427 on Build GRCh38
rs908742643 Chr.1: 1452521-1452521 on Build GRCh38
rs760502916 Chr.1: 1452395-1452395 on Build GRCh38
rs375847907 Chr.1: 1452441-1452441 on Build GRCh38
rs773954981 Chr.1: 1452447-1452447 on Build GRCh38
rs376231836 Chr.1: 1452397-1452397 on Build GRCh38
rs202238789 Chr.1: 1452323-1452323 on Build GRCh38
rs370314383 Chr.1: 1452414-1452414 on Build GRCh38
rs967309331 Chr.1: 1452401-1452401 on Build GRCh38
rs772657768 Chr.1: 1452458-1452458 on Build GRCh38
rs563943468 Chr.1: 1452318-1452318 on Build GRCh38
rs772942385 Chr.1: 1452390-1452390 on Build GRCh38
rs144852834 Chr.1: 1452396-1452396 on Build GRCh38
rs928208253 Chr.1: 1452515-1452515 on Build GRCh38
rs747573945 Chr.1: 1452456-1452456 on Build GRCh38
rs757248068 Chr.1: 1452434-1452434 on Build GRCh38
rs1624483 Chr.1: 1452511-1452511 on Build GRCh38
rs781079363 Chr.1: 1452388-1452388 on Build GRCh38
rs529649885 Chr.1: 1452319-1452319 on Build GRCh38
rs567033403 Chr.1: 1452389-1452389 on Build GRCh38
rs1049772544 Chr.1: 1452539-1452539 on Build GRCh38
rs527371033 Chr.1: 1452355-1452355 on Build GRCh38
rs557919320 Chr.1: 1452490-1452490 on Build GRCh38
rs776279430 Chr.1: 1452455-1452455 on Build GRCh38
rs753995263 Chr.1: 1452345-1452345 on Build GRCh38
rs377228484 Chr.1: 1452433-1452433 on Build GRCh38
rs534827134 Chr.1: 1452479-1452479 on Build GRCh38
rs930925740 Chr.1: 1452513-1452513 on Build GRCh38
rs200581700 Chr.1: 1452366-1452366 on Build GRCh38
rs759842500 Chr.1: 1452474-1452474 on Build GRCh38
rs535108799 Chr.1: 1452415-1452415 on Build GRCh38
rs1625156 Chr.1: 1452540-1452540 on Build GRCh38
rs1006677057 Chr.1: 1452369-1452369 on Build GRCh38
rs773798707 Chr.1: 1452464-1452464 on Build GRCh38
rs749002369 Chr.1: 1452446-1452446 on Build GRCh38
rs565775354 Chr.1: 1452477-1452477 on Build GRCh38
rs761125589 Chr.1: 1452471-1452471 on Build GRCh38
rs372549333 Chr.1: 1452421-1452421 on Build GRCh38
rs754048352 Chr.1: 1452423-1452423 on Build GRCh38
rs756446879 Chr.1: 1452340-1452340 on Build GRCh38
rs765958652 Chr.1: 1452440-1452445 on Build GRCh38

View Full Product Details