Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6400888-6401161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APBB1
Entrez Gene ID
322
Gene Name
amyloid beta precursor protein binding family B member 1
Gene Aliases
FE65, MGC:9072, RIR
UniGene
Hs.372840
Gene Chromosome Location
Chr.11: 6395124-6419830 on Build GRCh38
Transcripts
NM_001164.4 NM_001257319.2 NM_001257320.2 NM_001257321.2 NM_001257323.2 NM_001257325.2 NM_001257326.2 NM_145689.2 NR_047512.2 XM_006718209.3 XM_006718210.3 XM_006718211.1 XM_006718212.3 XM_006718213.1 XM_006718214.3 XM_006718215.3 XM_011520037.2 XM_011520038.2 XM_011520040.2 XM_011520042.1 XM_017017639.1 XM_017017640.1 XM_017017641.1 XM_017017642.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5839155 c.1666+1delG p.? Chr.11: 6400988-6400988 on Build GRCh38
50361 c.1654G>A p.A552T Chr.11: 6401001-6401001 on Build GRCh38
6601897 c.1592A>T p.E531V Chr.11: 6401069-6401069 on Build GRCh38
930123 c.1602G>T p.K534N Chr.11: 6401053-6401053 on Build GRCh38
1703509 c.1634A>T p.Y545F Chr.11: 6401021-6401021 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775551746 Chr.11: 6401104-6401104 on Build GRCh38
rs752575406 Chr.11: 6400974-6400974 on Build GRCh38
rs979700045 Chr.11: 6401007-6401007 on Build GRCh38
rs773670333 Chr.11: 6401057-6401057 on Build GRCh38
rs528578071 Chr.11: 6401035-6401035 on Build GRCh38
rs769451274 Chr.11: 6401042-6401042 on Build GRCh38
rs147235391 Chr.11: 6400995-6400995 on Build GRCh38
rs1012556601 Chr.11: 6400915-6400915 on Build GRCh38
rs975352359 Chr.11: 6400917-6400917 on Build GRCh38
rs947473389 Chr.11: 6400954-6400954 on Build GRCh38
rs990048714 Chr.11: 6401013-6401013 on Build GRCh38
rs755272809 Chr.11: 6401102-6401107 on Build GRCh38
rs879887643 Chr.11: 6401126-6401126 on Build GRCh38
rs897345240 Chr.11: 6401110-6401110 on Build GRCh38
rs114774881 Chr.11: 6401117-6401117 on Build GRCh38
rs759446538 Chr.11: 6401076-6401076 on Build GRCh38
rs777378458 Chr.11: 6401056-6401056 on Build GRCh38
rs771737627 Chr.11: 6401101-6401101 on Build GRCh38
rs370385427 Chr.11: 6400972-6400972 on Build GRCh38
rs949040469 Chr.11: 6401103-6401103 on Build GRCh38
rs754998524 Chr.11: 6401018-6401018 on Build GRCh38
rs1049683432 Chr.11: 6401082-6401082 on Build GRCh38
rs748954442 Chr.11: 6401059-6401059 on Build GRCh38
rs143081866 Chr.11: 6401011-6401011 on Build GRCh38
rs750854475 Chr.11: 6401020-6401020 on Build GRCh38
rs971513563 Chr.11: 6400955-6400955 on Build GRCh38
rs373363005 Chr.11: 6401039-6401039 on Build GRCh38
rs754412822 Chr.11: 6401087-6401089 on Build GRCh38
rs117210486 Chr.11: 6400929-6400929 on Build GRCh38
rs764159848 Chr.11: 6401121-6401121 on Build GRCh38
rs995246760 Chr.11: 6401125-6401125 on Build GRCh38
rs774129962 Chr.11: 6400950-6400950 on Build GRCh38
rs1027593894 Chr.11: 6401131-6401131 on Build GRCh38
rs764141140 Chr.11: 6401002-6401002 on Build GRCh38
rs376626035 Chr.11: 6401086-6401086 on Build GRCh38
rs767550833 Chr.11: 6400959-6400959 on Build GRCh38
rs140731057 Chr.11: 6401010-6401010 on Build GRCh38
rs779260081 Chr.11: 6401012-6401012 on Build GRCh38
rs770651554 Chr.11: 6401060-6401060 on Build GRCh38
rs267603097 Chr.11: 6401065-6401065 on Build GRCh38
rs773864621 Chr.11: 6401071-6401071 on Build GRCh38
rs201693590 Chr.11: 6401122-6401122 on Build GRCh38
rs375736918 Chr.11: 6400957-6400957 on Build GRCh38
rs766309407 Chr.11: 6400945-6400945 on Build GRCh38

View Full Product Details