Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 93994313-93994544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BET1
Entrez Gene ID
10282
Gene Name
Bet1 golgi vesicular membrane trafficking protein
Gene Aliases
HBET1
UniGene
Hs.489132
Gene Chromosome Location
Chr.7: 93962770-94004382 on Build GRCh38
Transcripts
NM_001317739.1 NM_005868.5 NR_133908.1 NR_133909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3883458 c.221C>A p.T74K Chr.7: 93994366-93994366 on Build GRCh38
1093242 c.202G>A p.D68N Chr.7: 93994385-93994385 on Build GRCh38
4778331 c.202-2A>G p.? Chr.7: 93994387-93994387 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768923163 Chr.7: 93994386-93994386 on Build GRCh38
rs1031333298 Chr.7: 93994417-93994417 on Build GRCh38
rs764744251 Chr.7: 93994412-93994412 on Build GRCh38
rs188078412 Chr.7: 93994517-93994517 on Build GRCh38
rs772338418 Chr.7: 93994359-93994359 on Build GRCh38
rs911911206 Chr.7: 93994437-93994437 on Build GRCh38
rs775800524 Chr.7: 93994360-93994360 on Build GRCh38
rs751358088 Chr.7: 93994364-93994364 on Build GRCh38
rs746270212 Chr.7: 93994496-93994497 on Build GRCh38
rs747449185 Chr.7: 93994376-93994376 on Build GRCh38
rs556839596 Chr.7: 93994366-93994366 on Build GRCh38
rs757059937 Chr.7: 93994414-93994415 on Build GRCh38
rs542472377 Chr.7: 93994496-93994496 on Build GRCh38
rs762228549 Chr.7: 93994394-93994394 on Build GRCh38
rs997136566 Chr.7: 93994396-93994396 on Build GRCh38
rs746032027 Chr.7: 93994355-93994355 on Build GRCh38
rs777155325 Chr.7: 93994388-93994388 on Build GRCh38
rs573571287 Chr.7: 93994458-93994458 on Build GRCh38
rs891703761 Chr.7: 93994455-93994455 on Build GRCh38
rs750197326 Chr.7: 93994438-93994438 on Build GRCh38
rs774778346 Chr.7: 93994392-93994392 on Build GRCh38
rs1047703776 Chr.7: 93994436-93994436 on Build GRCh38

View Full Product Details