Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 34712678-34712951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFL2
Entrez Gene ID
1073
Gene Name
cofilin 2
Gene Aliases
NEM7
UniGene
Hs.180141
Gene Chromosome Location
Chr.14: 34710382-34714823 on Build GRCh38
Transcripts
NM_001243645.1 NM_021914.7 NM_138638.4 NR_028130.1 NR_028131.1 XM_011536363.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1742999 c.470T>A p.V157E Chr.14: 34712896-34712896 on Build GRCh38
6140648 c.461G>C p.G154A Chr.14: 34712905-34712905 on Build GRCh38
6042775 c.443C>T p.T148I Chr.14: 34712923-34712923 on Build GRCh38
6140647 c.461G>C p.G154A Chr.14: 34712905-34712905 on Build GRCh38
4548979 c.461G>A p.G154E Chr.14: 34712905-34712905 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540447803 Chr.14: 34712853-34712853 on Build GRCh38
rs758416058 Chr.14: 34712739-34712739 on Build GRCh38
rs201619073 Chr.14: 34712871-34712871 on Build GRCh38
rs752855805 Chr.14: 34712745-34712745 on Build GRCh38
rs757466974 Chr.14: 34712782-34712782 on Build GRCh38
rs932113690 Chr.14: 34712916-34712916 on Build GRCh38
rs757526740 Chr.14: 34712926-34712926 on Build GRCh38
rs766364990 Chr.14: 34712874-34712874 on Build GRCh38
rs775818350 Chr.14: 34712815-34712815 on Build GRCh38
rs749446481 Chr.14: 34712780-34712780 on Build GRCh38
rs904019612 Chr.14: 34712834-34712834 on Build GRCh38
rs879179615 Chr.14: 34712858-34712858 on Build GRCh38
rs750236103 Chr.14: 34712725-34712725 on Build GRCh38
rs547962200 Chr.14: 34712890-34712890 on Build GRCh38
rs74043541 Chr.14: 34712825-34712825 on Build GRCh38
rs916778894 Chr.14: 34712798-34712798 on Build GRCh38
rs145938782 Chr.14: 34712925-34712925 on Build GRCh38
rs773187024 Chr.14: 34712839-34712839 on Build GRCh38
rs531587859 Chr.14: 34712821-34712821 on Build GRCh38
rs749975663 Chr.14: 34712748-34712748 on Build GRCh38
rs751399023 Chr.14: 34712885-34712885 on Build GRCh38
rs983181805 Chr.14: 34712704-34712704 on Build GRCh38
rs777987973 Chr.14: 34712767-34712767 on Build GRCh38
rs746198402 Chr.14: 34712790-34712790 on Build GRCh38
rs563036298 Chr.14: 34712740-34712740 on Build GRCh38
rs115050412 Chr.14: 34712830-34712830 on Build GRCh38
rs779309520 Chr.14: 34712785-34712785 on Build GRCh38
rs753788777 Chr.14: 34712925-34712926 on Build GRCh38
rs776423409 Chr.14: 34712829-34712829 on Build GRCh38
rs587780312 Chr.14: 34712909-34712909 on Build GRCh38
rs756102382 Chr.14: 34712757-34712757 on Build GRCh38
rs772673889 Chr.14: 34712810-34712810 on Build GRCh38

View Full Product Details