Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 14752657-14752926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD30B
Entrez Gene ID
374860
Gene Name
ankyrin repeat domain 30B
Gene Aliases
NY-BR-1.1
UniGene
Hs.567889
Gene Chromosome Location
Chr.18: 14748240-14855465 on Build GRCh38
Transcripts
NM_001145029.1 XM_006722325.3 XM_011525662.2 XM_011525664.2 XM_011525665.2 XM_011525666.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6453777 c.326C>G p.P109R Chr.18: 14752670-14752670 on Build GRCh38
6453778 c.326C>G p.P109R Chr.18: 14752670-14752670 on Build GRCh38
3524360 c.336G>A p.K112K Chr.18: 14752680-14752680 on Build GRCh38
3937694 c.406G>C p.D136H Chr.18: 14752908-14752908 on Build GRCh38
5515663 c.336+8G>T p.? Chr.18: 14752688-14752688 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760214190 Chr.18: 14752820-14752820 on Build GRCh38
rs12959778 Chr.18: 14752724-14752724 on Build GRCh38
rs572652697 Chr.18: 14752855-14752855 on Build GRCh38
rs556937555 Chr.18: 14752807-14752807 on Build GRCh38
rs747990388 Chr.18: 14752896-14752896 on Build GRCh38
rs759791878 Chr.18: 14752711-14752711 on Build GRCh38
rs756853799 Chr.18: 14752864-14752864 on Build GRCh38
rs374547236 Chr.18: 14752817-14752817 on Build GRCh38
rs576848402 Chr.18: 14752774-14752774 on Build GRCh38
rs988122916 Chr.18: 14752737-14752737 on Build GRCh38
rs574383616 Chr.18: 14752705-14752705 on Build GRCh38
rs939352410 Chr.18: 14752721-14752721 on Build GRCh38
rs757471506 Chr.18: 14752883-14752883 on Build GRCh38
rs559142364 Chr.18: 14752851-14752851 on Build GRCh38
rs771104933 Chr.18: 14752706-14752706 on Build GRCh38
rs753488886 Chr.18: 14752832-14752832 on Build GRCh38
rs771819072 Chr.18: 14752900-14752900 on Build GRCh38
rs768351086 Chr.18: 14752727-14752727 on Build GRCh38
rs199597228 Chr.18: 14752850-14752850 on Build GRCh38
rs746084131 Chr.18: 14752884-14752884 on Build GRCh38
rs774040196 Chr.18: 14752892-14752892 on Build GRCh38
rs747513542 Chr.18: 14752875-14752875 on Build GRCh38
rs1039427827 Chr.18: 14752790-14752790 on Build GRCh38
rs767398172 Chr.18: 14752814-14752814 on Build GRCh38
rs1001858384 Chr.18: 14752771-14752771 on Build GRCh38
rs45487996 Chr.18: 14752740-14752740 on Build GRCh38
rs774131799 Chr.18: 14752805-14752805 on Build GRCh38
rs1042961820 Chr.18: 14752742-14752742 on Build GRCh38
rs913146746 Chr.18: 14752738-14752738 on Build GRCh38
rs900948445 Chr.18: 14752791-14752791 on Build GRCh38
rs748614523 Chr.18: 14752690-14752690 on Build GRCh38
rs772638879 Chr.18: 14752693-14752693 on Build GRCh38
rs942545723 Chr.18: 14752786-14752786 on Build GRCh38
rs777383479 Chr.18: 14752746-14752746 on Build GRCh38
rs778472408 Chr.18: 14752872-14752872 on Build GRCh38
rs776785994 Chr.18: 14752707-14752707 on Build GRCh38
rs765905553 Chr.18: 14752825-14752825 on Build GRCh38
rs755361447 Chr.18: 14752685-14752685 on Build GRCh38
rs779320927 Chr.18: 14752686-14752686 on Build GRCh38
rs45620343 Chr.18: 14752769-14752769 on Build GRCh38
rs770021117 Chr.18: 14752885-14752885 on Build GRCh38

View Full Product Details