Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 89591117-89591388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2B1
Entrez Gene ID
490
Gene Name
ATPase plasma membrane Ca2+ transporting 1
Gene Aliases
PMCA1, PMCA1kb
UniGene
Hs.506276
Gene Chromosome Location
Chr.12: 89588049-89709353 on Build GRCh38
Transcripts
NM_001001323.1 NM_001682.2 XM_011538407.2 XM_017019357.1 XM_017019358.1 XM_017019359.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
274090 c.3419C>T p.S1140L Chr.12: 89591228-89591228 on Build GRCh38
3466281 c.3413G>A p.R1138K Chr.12: 89591234-89591234 on Build GRCh38
6505314 c.3392G>A p.G1131E Chr.12: 89591255-89591255 on Build GRCh38
3466282 c.3373C>T p.R1125C Chr.12: 89591274-89591274 on Build GRCh38
3955278 c.3352-1G>C p.? Chr.12: 89591296-89591296 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369985301 Chr.12: 89591242-89591242 on Build GRCh38
rs375435006 Chr.12: 89591192-89591192 on Build GRCh38
rs777051043 Chr.12: 89591189-89591189 on Build GRCh38
rs757086886 Chr.12: 89591354-89591354 on Build GRCh38
rs576521274 Chr.12: 89591329-89591329 on Build GRCh38
rs746568307 Chr.12: 89591302-89591302 on Build GRCh38
rs762207543 Chr.12: 89591193-89591193 on Build GRCh38
rs753652453 Chr.12: 89591231-89591231 on Build GRCh38
rs922455554 Chr.12: 89591259-89591259 on Build GRCh38
rs775232867 Chr.12: 89591211-89591211 on Build GRCh38
rs764946874 Chr.12: 89591243-89591243 on Build GRCh38
rs769105123 Chr.12: 89591174-89591174 on Build GRCh38
rs760644092 Chr.12: 89591219-89591219 on Build GRCh38
rs61736878 Chr.12: 89591227-89591227 on Build GRCh38
rs762977872 Chr.12: 89591203-89591203 on Build GRCh38
rs766483886 Chr.12: 89591205-89591205 on Build GRCh38
rs750107374 Chr.12: 89591263-89591263 on Build GRCh38
rs745346247 Chr.12: 89591145-89591145 on Build GRCh38
rs761652063 Chr.12: 89591338-89591340 on Build GRCh38
rs776669934 Chr.12: 89591322-89591323 on Build GRCh38
rs373795130 Chr.12: 89591173-89591173 on Build GRCh38
rs945772727 Chr.12: 89591149-89591149 on Build GRCh38
rs769982978 Chr.12: 89591195-89591195 on Build GRCh38
rs781599299 Chr.12: 89591332-89591332 on Build GRCh38
rs751758239 Chr.12: 89591197-89591197 on Build GRCh38
rs770173640 Chr.12: 89591344-89591344 on Build GRCh38
rs369043635 Chr.12: 89591147-89591147 on Build GRCh38
rs753819762 Chr.12: 89591340-89591340 on Build GRCh38
rs771742877 Chr.12: 89591146-89591146 on Build GRCh38
rs181715279 Chr.12: 89591299-89591299 on Build GRCh38

View Full Product Details