Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7486554-7486809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD163
Entrez Gene ID
9332
Gene Name
CD163 molecule
Gene Aliases
M130, MM130, SCARI1
UniGene
Hs.504641
Gene Chromosome Location
Chr.12: 7470816-7503818 on Build GRCh38
Transcripts
NM_004244.5 NM_203416.3 XM_005253528.4 XM_005253529.4 XM_017020231.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7209406 c.2480G>A p.R827H Chr.12: 7486576-7486576 on Build GRCh38
7209405 c.2381G>A p.R794H Chr.12: 7486576-7486576 on Build GRCh38
5603114 c.2321G>A p.G774D Chr.12: 7486735-7486735 on Build GRCh38
3465405 c.2477C>T p.S826F Chr.12: 7486579-7486579 on Build GRCh38
4044861 c.2389G>T p.E797* Chr.12: 7486667-7486667 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs916825435 Chr.12: 7486626-7486626 on Build GRCh38
rs749980745 Chr.12: 7486694-7486694 on Build GRCh38
rs747400028 Chr.12: 7486778-7486778 on Build GRCh38
rs981079611 Chr.12: 7486664-7486665 on Build GRCh38
rs776564335 Chr.12: 7486685-7486685 on Build GRCh38
rs761562814 Chr.12: 7486692-7486692 on Build GRCh38
rs776582425 Chr.12: 7486780-7486780 on Build GRCh38
rs201192099 Chr.12: 7486696-7486696 on Build GRCh38
rs758460376 Chr.12: 7486757-7486757 on Build GRCh38
rs746074392 Chr.12: 7486746-7486746 on Build GRCh38
rs970048297 Chr.12: 7486620-7486620 on Build GRCh38
rs374511072 Chr.12: 7486591-7486591 on Build GRCh38
rs375516469 Chr.12: 7486624-7486624 on Build GRCh38
rs919570406 Chr.12: 7486667-7486667 on Build GRCh38
rs771070907 Chr.12: 7486603-7486603 on Build GRCh38
rs768631645 Chr.12: 7486674-7486674 on Build GRCh38
rs1040428201 Chr.12: 7486743-7486743 on Build GRCh38
rs775333514 Chr.12: 7486648-7486648 on Build GRCh38
rs756517724 Chr.12: 7486577-7486577 on Build GRCh38
rs778342413 Chr.12: 7486741-7486741 on Build GRCh38
rs778112907 Chr.12: 7486717-7486717 on Build GRCh38
rs768647918 Chr.12: 7486777-7486777 on Build GRCh38
rs760670850 Chr.12: 7486662-7486662 on Build GRCh38
rs772081770 Chr.12: 7486646-7486646 on Build GRCh38
rs373948816 Chr.12: 7486695-7486695 on Build GRCh38
rs762219071 Chr.12: 7486576-7486576 on Build GRCh38
rs1040900739 Chr.12: 7486729-7486729 on Build GRCh38
rs747012462 Chr.12: 7486776-7486776 on Build GRCh38
rs778094768 Chr.12: 7486588-7486588 on Build GRCh38
rs918143071 Chr.12: 7486595-7486595 on Build GRCh38
rs752244536 Chr.12: 7486783-7486783 on Build GRCh38
rs147727599 Chr.12: 7486719-7486719 on Build GRCh38
rs756607423 Chr.12: 7486698-7486698 on Build GRCh38
rs780170049 Chr.12: 7486773-7486773 on Build GRCh38
rs779060172 Chr.12: 7486616-7486616 on Build GRCh38
rs757497545 Chr.12: 7486734-7486734 on Build GRCh38

View Full Product Details