Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137048986-137049235 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD2
Entrez Gene ID
954
Gene Name
ectonucleoside triphosphate diphosphohydrolase 2
Gene Aliases
CD39L1, NTPDase-2
UniGene
Hs.123036
Gene Chromosome Location
Chr.9: 137048098-137054051 on Build GRCh38
Transcripts
NM_001246.3 NM_203468.2 XM_011519212.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5977163 c.1179C>T p.A393A Chr.9: 137049046-137049046 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs7390981 Chr.9: 137049097-137049097 on Build GRCh38
rs778301758 Chr.9: 137049092-137049092 on Build GRCh38
rs774359077 Chr.9: 137049171-137049171 on Build GRCh38
rs554966421 Chr.9: 137049146-137049146 on Build GRCh38
rs750143962 Chr.9: 137049133-137049133 on Build GRCh38
rs553845490 Chr.9: 137049214-137049214 on Build GRCh38
rs536251205 Chr.9: 137049211-137049211 on Build GRCh38
rs771657346 Chr.9: 137049007-137049007 on Build GRCh38
rs766679372 Chr.9: 137049215-137049215 on Build GRCh38
rs948706107 Chr.9: 137049163-137049163 on Build GRCh38
rs755466572 Chr.9: 137049090-137049090 on Build GRCh38
rs777952982 Chr.9: 137049185-137049185 on Build GRCh38
rs756528638 Chr.9: 137049087-137049087 on Build GRCh38
rs368497640 Chr.9: 137049132-137049132 on Build GRCh38
rs749794737 Chr.9: 137049089-137049089 on Build GRCh38
rs1048332609 Chr.9: 137049041-137049041 on Build GRCh38
rs907248514 Chr.9: 137049028-137049028 on Build GRCh38
rs45457700 Chr.9: 137049085-137049085 on Build GRCh38
rs540334692 Chr.9: 137049209-137049209 on Build GRCh38
rs374463317 Chr.9: 137049046-137049046 on Build GRCh38
rs751597294 Chr.9: 137049065-137049065 on Build GRCh38
rs914494484 Chr.9: 137049162-137049162 on Build GRCh38
rs759404134 Chr.9: 137049166-137049166 on Build GRCh38
rs777391003 Chr.9: 137049107-137049107 on Build GRCh38
rs761967508 Chr.9: 137049191-137049191 on Build GRCh38
rs955088455 Chr.9: 137049122-137049122 on Build GRCh38
rs868390289 Chr.9: 137049131-137049131 on Build GRCh38
rs1028032104 Chr.9: 137049115-137049115 on Build GRCh38
rs770743751 Chr.9: 137049145-137049145 on Build GRCh38
rs746846153 Chr.9: 137049108-137049108 on Build GRCh38
rs758258946 Chr.9: 137049125-137049125 on Build GRCh38
rs766353234 Chr.9: 137049036-137049036 on Build GRCh38
rs938725160 Chr.9: 137049184-137049184 on Build GRCh38
rs558929657 Chr.9: 137049017-137049017 on Build GRCh38
rs924415207 Chr.9: 137049143-137049143 on Build GRCh38
rs990508783 Chr.9: 137049152-137049152 on Build GRCh38
rs1037477877 Chr.9: 137049201-137049201 on Build GRCh38
rs757422958 Chr.9: 137049084-137049084 on Build GRCh38
rs778810351 Chr.9: 137049043-137049043 on Build GRCh38
rs975682620 Chr.9: 137049095-137049095 on Build GRCh38
rs760459410 Chr.9: 137049033-137049033 on Build GRCh38
rs771780022 Chr.9: 137049106-137049106 on Build GRCh38
rs371094769 Chr.9: 137049050-137049050 on Build GRCh38
rs765013535 Chr.9: 137049027-137049049 on Build GRCh38
rs989553593 Chr.9: 137049039-137049039 on Build GRCh38
rs569953168 Chr.9: 137049204-137049204 on Build GRCh38
rs752603291 Chr.9: 137049048-137049048 on Build GRCh38
rs1031060996 Chr.9: 137049018-137049018 on Build GRCh38
rs1003052514 Chr.9: 137049029-137049029 on Build GRCh38
rs781618182 Chr.9: 137049149-137049150 on Build GRCh38
rs780751115 Chr.9: 137049088-137049088 on Build GRCh38

View Full Product Details