Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 63789029-63789254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EYS
Entrez Gene ID
346007
Gene Name
eyes shut homolog (Drosophila)
Gene Aliases
C6orf178, C6orf179, C6orf180, EGFL10, EGFL11, RP25, SPAM, bA166P24.2, bA307F22.3, bA74E24.1, dJ1018A4.2, dJ22I17.2, dJ303F19.1
UniGene
Hs.25067
Gene Chromosome Location
Chr.6: 63719980-65707225 on Build GRCh38
Transcripts
NM_001142800.1 NM_001292009.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1445570 c.7453G>T p.G2485C Chr.6: 63789183-63789183 on Build GRCh38
1445569 c.7510C>A p.P2504T Chr.6: 63789126-63789126 on Build GRCh38
4625829 c.7547T>C p.L2516P Chr.6: 63789089-63789089 on Build GRCh38
1445568 c.7566G>A p.E2522E Chr.6: 63789070-63789070 on Build GRCh38
6842776 c.7412-1G>C p.? Chr.6: 63789225-63789225 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759150246 Chr.6: 63789179-63789179 on Build GRCh38
rs774918307 Chr.6: 63789195-63789195 on Build GRCh38
rs1044722026 Chr.6: 63789193-63789193 on Build GRCh38
rs1013443458 Chr.6: 63789174-63789174 on Build GRCh38
rs560012389 Chr.6: 63789182-63789182 on Build GRCh38
rs954205055 Chr.6: 63789125-63789125 on Build GRCh38
rs902462590 Chr.6: 63789064-63789064 on Build GRCh38
rs140594243 Chr.6: 63789213-63789213 on Build GRCh38
rs760224932 Chr.6: 63789214-63789214 on Build GRCh38
rs1020797365 Chr.6: 63789051-63789051 on Build GRCh38
rs527486914 Chr.6: 63789078-63789078 on Build GRCh38
rs768964978 Chr.6: 63789129-63789129 on Build GRCh38
rs77113519 Chr.6: 63789130-63789130 on Build GRCh38
rs199754359 Chr.6: 63789087-63789087 on Build GRCh38
rs781576158 Chr.6: 63789170-63789170 on Build GRCh38
rs528819037 Chr.6: 63789067-63789067 on Build GRCh38

View Full Product Details