Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 111244104-111244353 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF7
Entrez Gene ID
8874
Gene Name
Rho guanine nucleotide exchange factor 7
Gene Aliases
BETA-PIX, COOL-1, COOL1, Nbla10314, P50, P50BP, P85, P85COOL1, P85SPR, PAK3, PIXB
UniGene
Hs.508738
Gene Chromosome Location
Chr.13: 111114619-111305734 on Build GRCh38
Transcripts
NM_001113511.2 NM_001113512.2 NM_001113513.1 NM_001320851.1 NM_001320852.1 NM_001320853.1 NM_001320854.1 NM_003899.3 NM_145735.3 XM_006719956.3 XM_011521133.2 XM_017020814.1 XM_017020815.1 XM_017020816.1 XM_017020817.1 XM_017020818.1 XM_017020819.1 XM_017020820.1 XM_017020821.1 XM_017020822.1 XM_017020823.1 XM_017020824.1 XM_017020825.1 XM_017020826.1 XM_017020827.1 XM_017020828.1 XM_017020829.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1660240 c.392C>T p.S131L Chr.13: 111244207-111244207 on Build GRCh38
1365502 c.940G>A p.E314K Chr.13: 111244284-111244284 on Build GRCh38
6000925 c.928G>A p.A310T Chr.13: 111244209-111244209 on Build GRCh38
5065993 c.961G>A p.E321K Chr.13: 111244242-111244242 on Build GRCh38
1562373 c.427G>A p.E143K Chr.13: 111244242-111244242 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs930935192 Chr.13: 111244306-111244306 on Build GRCh38
rs150270871 Chr.13: 111244220-111244220 on Build GRCh38
rs764473169 Chr.13: 111244283-111244283 on Build GRCh38
rs928499638 Chr.13: 111244146-111244146 on Build GRCh38
rs751798965 Chr.13: 111244152-111244152 on Build GRCh38
rs767688172 Chr.13: 111244268-111244268 on Build GRCh38
rs747945936 Chr.13: 111244221-111244221 on Build GRCh38
rs61742189 Chr.13: 111244290-111244290 on Build GRCh38
rs756485963 Chr.13: 111244177-111244177 on Build GRCh38
rs757772711 Chr.13: 111244294-111244294 on Build GRCh38
rs938406644 Chr.13: 111244157-111244157 on Build GRCh38
rs376300554 Chr.13: 111244160-111244160 on Build GRCh38
rs755131869 Chr.13: 111244159-111244159 on Build GRCh38
rs753069956 Chr.13: 111244164-111244164 on Build GRCh38
rs752939884 Chr.13: 111244269-111244269 on Build GRCh38
rs779562617 Chr.13: 111244304-111244304 on Build GRCh38
rs773161913 Chr.13: 111244249-111244249 on Build GRCh38
rs769523142 Chr.13: 111244245-111244245 on Build GRCh38
rs759607285 Chr.13: 111244266-111244266 on Build GRCh38
rs182815658 Chr.13: 111244259-111244259 on Build GRCh38
rs754359059 Chr.13: 111244203-111244203 on Build GRCh38
rs768264835 Chr.13: 111244215-111244215 on Build GRCh38
rs751176796 Chr.13: 111244305-111244305 on Build GRCh38
rs746665081 Chr.13: 111244214-111244214 on Build GRCh38
rs761596415 Chr.13: 111244287-111244287 on Build GRCh38
rs760978338 Chr.13: 111244281-111244281 on Build GRCh38
rs762823926 Chr.13: 111244255-111244255 on Build GRCh38
rs778191238 Chr.13: 111244192-111244192 on Build GRCh38
rs774347347 Chr.13: 111244264-111244264 on Build GRCh38
rs1048541537 Chr.13: 111244312-111244312 on Build GRCh38
rs758005923 Chr.13: 111244206-111244206 on Build GRCh38
rs200372907 Chr.13: 111244229-111244229 on Build GRCh38

View Full Product Details