Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18008968-18009209 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARRDC2
Entrez Gene ID
27106
Gene Name
arrestin domain containing 2
Gene Aliases
CLONE24945, PP2703
UniGene
Hs.515249
Gene Chromosome Location
Chr.19: 18001132-18014102 on Build GRCh38
Transcripts
NM_001025604.2 NM_001286826.1 NM_015683.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6003829 c.455T>C p.I152T Chr.19: 18009084-18009084 on Build GRCh38
374292 c.412C>T p.R138C Chr.19: 18009056-18009056 on Build GRCh38
6605232 c.358G>A p.G120S Chr.19: 18009002-18009002 on Build GRCh38
5972797 c.453C>T p.V151V Chr.19: 18009082-18009082 on Build GRCh38
6999098 c.342-2A>G p.? Chr.19: 18008969-18008969 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200754201 Chr.19: 18009111-18009111 on Build GRCh38
rs983038115 Chr.19: 18009143-18009143 on Build GRCh38
rs755564060 Chr.19: 18009164-18009164 on Build GRCh38
rs201158191 Chr.19: 18009060-18009060 on Build GRCh38
rs774298500 Chr.19: 18009120-18009120 on Build GRCh38
rs143985791 Chr.19: 18009049-18009049 on Build GRCh38
rs981350891 Chr.19: 18009183-18009183 on Build GRCh38
rs751434752 Chr.19: 18009065-18009065 on Build GRCh38
rs12610718 Chr.19: 18009032-18009032 on Build GRCh38
rs749725166 Chr.19: 18009086-18009086 on Build GRCh38
rs748628072 Chr.19: 18009095-18009095 on Build GRCh38
rs756733526 Chr.19: 18009069-18009069 on Build GRCh38
rs201677116 Chr.19: 18009040-18009040 on Build GRCh38
rs146041752 Chr.19: 18009061-18009061 on Build GRCh38
rs374193417 Chr.19: 18009105-18009105 on Build GRCh38
rs150741664 Chr.19: 18009037-18009037 on Build GRCh38
rs756789716 Chr.19: 18009002-18009002 on Build GRCh38
rs547129702 Chr.19: 18009056-18009056 on Build GRCh38
rs780715164 Chr.19: 18009011-18009011 on Build GRCh38
rs771201998 Chr.19: 18009108-18009108 on Build GRCh38
rs767008804 Chr.19: 18009070-18009070 on Build GRCh38
rs569309020 Chr.19: 18009184-18009184 on Build GRCh38
rs766606529 Chr.19: 18009130-18009130 on Build GRCh38
rs749995145 Chr.19: 18009152-18009152 on Build GRCh38
rs763821805 Chr.19: 18009118-18009118 on Build GRCh38
rs776830538 Chr.19: 18009110-18009110 on Build GRCh38
rs368697056 Chr.19: 18009185-18009185 on Build GRCh38
rs762049398 Chr.19: 18009047-18009047 on Build GRCh38
rs755386468 Chr.19: 18009088-18009088 on Build GRCh38
rs747038483 Chr.19: 18009106-18009106 on Build GRCh38
rs767322430 Chr.19: 18009150-18009150 on Build GRCh38
rs774982292 Chr.19: 18009042-18009042 on Build GRCh38
rs750493307 Chr.19: 18009124-18009124 on Build GRCh38
rs767857478 Chr.19: 18009059-18009059 on Build GRCh38
rs779566593 Chr.19: 18009089-18009089 on Build GRCh38
rs189196235 Chr.19: 18008992-18008992 on Build GRCh38
rs12610716 Chr.19: 18009013-18009013 on Build GRCh38
rs371134871 Chr.19: 18009038-18009038 on Build GRCh38
rs751130030 Chr.19: 18009001-18009001 on Build GRCh38
rs779826816 Chr.19: 18009085-18009085 on Build GRCh38
rs965673407 Chr.19: 18009155-18009155 on Build GRCh38
rs761774256 Chr.19: 18009136-18009136 on Build GRCh38
rs772066786 Chr.19: 18009100-18009100 on Build GRCh38
rs750077643 Chr.19: 18009081-18009081 on Build GRCh38
rs904397117 Chr.19: 18009007-18009007 on Build GRCh38
rs755826943 Chr.19: 18009082-18009082 on Build GRCh38
rs1046806 Chr.19: 18009039-18009039 on Build GRCh38
rs76941687 Chr.19: 18009012-18009012 on Build GRCh38
rs563100760 Chr.19: 18009090-18009090 on Build GRCh38

View Full Product Details