Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 30743995-30744240 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOX5AP
Entrez Gene ID
241
Gene Name
arachidonate 5-lipoxygenase activating protein
Gene Aliases
FLAP
UniGene
Hs.507658
Gene Chromosome Location
Chr.13: 30713478-30764428 on Build GRCh38
Transcripts
NM_001204406.1 NM_001629.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5433270 c.95A>T p.H32L Chr.13: 30744084-30744084 on Build GRCh38
278792 c.75C>A p.F25L Chr.13: 30744064-30744064 on Build GRCh38
946637 c.80C>T p.A27V Chr.13: 30744069-30744069 on Build GRCh38
3885224 c.120G>A p.R40R Chr.13: 30744109-30744109 on Build GRCh38
3671262 c.71G>T p.G24V Chr.13: 30744060-30744060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201221950 Chr.13: 30744209-30744209 on Build GRCh38
rs761318584 Chr.13: 30744094-30744094 on Build GRCh38
rs375032484 Chr.13: 30744077-30744077 on Build GRCh38
rs755960673 Chr.13: 30744204-30744204 on Build GRCh38
rs3803277 Chr.13: 30744171-30744171 on Build GRCh38
rs776630925 Chr.13: 30744064-30744064 on Build GRCh38
rs751043129 Chr.13: 30744092-30744092 on Build GRCh38
rs781044231 Chr.13: 30744156-30744156 on Build GRCh38
rs1038621588 Chr.13: 30744056-30744056 on Build GRCh38
rs201830648 Chr.13: 30744144-30744144 on Build GRCh38
rs752662748 Chr.13: 30744203-30744203 on Build GRCh38
rs989876575 Chr.13: 30744184-30744184 on Build GRCh38
rs200113122 Chr.13: 30744164-30744164 on Build GRCh38
rs978215931 Chr.13: 30744205-30744205 on Build GRCh38
rs756591705 Chr.13: 30744120-30744120 on Build GRCh38
rs372030223 Chr.13: 30744124-30744124 on Build GRCh38
rs780477456 Chr.13: 30744032-30744032 on Build GRCh38
rs756542676 Chr.13: 30744022-30744022 on Build GRCh38
rs778888248 Chr.13: 30744037-30744037 on Build GRCh38
rs201495068 Chr.13: 30744050-30744050 on Build GRCh38
rs767027375 Chr.13: 30744210-30744210 on Build GRCh38
rs41351946 Chr.13: 30744112-30744112 on Build GRCh38
rs141376946 Chr.13: 30744085-30744085 on Build GRCh38
rs199818248 Chr.13: 30744031-30744031 on Build GRCh38
rs200490573 Chr.13: 30744044-30744044 on Build GRCh38
rs143706085 Chr.13: 30744143-30744143 on Build GRCh38
rs201847785 Chr.13: 30744097-30744097 on Build GRCh38
rs202068154 Chr.13: 30744203-30744203 on Build GRCh38
rs768197433 Chr.13: 30744198-30744198 on Build GRCh38
rs772183678 Chr.13: 30744153-30744153 on Build GRCh38
rs775287884 Chr.13: 30744185-30744185 on Build GRCh38
rs911872602 Chr.13: 30744176-30744176 on Build GRCh38
rs960041029 Chr.13: 30744049-30744049 on Build GRCh38
rs759312162 Chr.13: 30744074-30744074 on Build GRCh38
rs201168290 Chr.13: 30744028-30744028 on Build GRCh38
rs763689154 Chr.13: 30744090-30744090 on Build GRCh38
rs748246562 Chr.13: 30744150-30744150 on Build GRCh38
rs749495238 Chr.13: 30744033-30744033 on Build GRCh38
rs767974846 Chr.13: 30744095-30744095 on Build GRCh38
rs201018097 Chr.13: 30744207-30744207 on Build GRCh38
rs762676655 Chr.13: 30744193-30744193 on Build GRCh38
rs374627262 Chr.13: 30744035-30744035 on Build GRCh38
rs146973182 Chr.13: 30744086-30744086 on Build GRCh38

View Full Product Details