Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150796676-150796937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSK
Entrez Gene ID
1513
Gene Name
cathepsin K
Gene Aliases
CTS02, CTSO, CTSO1, CTSO2, PKND, PYCD
UniGene
Hs.632466
Gene Chromosome Location
Chr.1: 150796208-150808441 on Build GRCh38
Transcripts
NM_000396.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4022048 c.990A>G p.*330W Chr.1: 150796799-150796799 on Build GRCh38
4382242 c.949G>A p.A317T Chr.1: 150796840-150796840 on Build GRCh38
6503720 c.895G>A p.G299R Chr.1: 150796894-150796894 on Build GRCh38
6898440 c.984G>A p.K328K Chr.1: 150796805-150796805 on Build GRCh38
4431122 c.935G>A p.R312Q Chr.1: 150796854-150796854 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1040117244 Chr.1: 150796741-150796741 on Build GRCh38
rs1025182609 Chr.1: 150796798-150796798 on Build GRCh38
rs201817083 Chr.1: 150796804-150796804 on Build GRCh38
rs774969011 Chr.1: 150796740-150796740 on Build GRCh38
rs141482674 Chr.1: 150796841-150796841 on Build GRCh38
rs756250449 Chr.1: 150796881-150796881 on Build GRCh38
rs199882930 Chr.1: 150796851-150796851 on Build GRCh38
rs199660923 Chr.1: 150796785-150796785 on Build GRCh38
rs773116784 Chr.1: 150796834-150796834 on Build GRCh38
rs768142570 Chr.1: 150796756-150796756 on Build GRCh38
rs956881073 Chr.1: 150796748-150796748 on Build GRCh38
rs761159349 Chr.1: 150796802-150796802 on Build GRCh38
rs1049965775 Chr.1: 150796718-150796718 on Build GRCh38
rs74315301 Chr.1: 150796799-150796799 on Build GRCh38
rs29001685 Chr.1: 150796863-150796863 on Build GRCh38
rs763968223 Chr.1: 150796874-150796874 on Build GRCh38
rs765168635 Chr.1: 150796826-150796826 on Build GRCh38
rs587756638 Chr.1: 150796840-150796840 on Build GRCh38
rs778490830 Chr.1: 150796888-150796888 on Build GRCh38
rs760596117 Chr.1: 150796841-150796843 on Build GRCh38
rs375958814 Chr.1: 150796855-150796855 on Build GRCh38
rs587638055 Chr.1: 150796864-150796864 on Build GRCh38
rs200118513 Chr.1: 150796806-150796806 on Build GRCh38
rs201185711 Chr.1: 150796757-150796757 on Build GRCh38
rs752348151 Chr.1: 150796805-150796805 on Build GRCh38
rs200655276 Chr.1: 150796796-150796796 on Build GRCh38
rs757744218 Chr.1: 150796896-150796896 on Build GRCh38
rs762780994 Chr.1: 150796836-150796836 on Build GRCh38
rs201702611 Chr.1: 150796849-150796849 on Build GRCh38
rs200704218 Chr.1: 150796721-150796721 on Build GRCh38

View Full Product Details