Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39021417-39021690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOBEC3D
Entrez Gene ID
140564
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3D
Gene Aliases
A3D, APOBEC3DE, APOBEC3E, ARP6
UniGene
Hs.629775
Gene Chromosome Location
Chr.22: 39021113-39034328 on Build GRCh38
Transcripts
NM_152426.3 XM_005261337.3 XM_017028596.1 XM_017028643.1

Gene Symbol
APOBEC3F
Entrez Gene ID
200316
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3F
Gene Aliases
A3F, ARP8, BK150C2.4.MRNA, KA6
UniGene
Hs.659809
Gene Chromosome Location
Chr.22: 39020454-39055972 on Build GRCh38
Transcripts
NM_152426.3 XM_005261337.3 XM_017028596.1 XM_017028643.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6875686 c.3G>A p.M1I Chr.22: 39021522-39021522 on Build GRCh38
461090 c.15C>A p.I5I Chr.22: 39021534-39021534 on Build GRCh38
5594199 c.15C>T p.I5I Chr.22: 39021534-39021534 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs374324581 Chr.22: 39021571-39021571 on Build GRCh38
rs944198906 Chr.22: 39021467-39021467 on Build GRCh38
rs760595347 Chr.22: 39021475-39021476 on Build GRCh38
rs748983584 Chr.22: 39021565-39021565 on Build GRCh38
rs987237370 Chr.22: 39021446-39021446 on Build GRCh38
rs200759595 Chr.22: 39021542-39021542 on Build GRCh38
rs532188755 Chr.22: 39021631-39021631 on Build GRCh38
rs906131715 Chr.22: 39021670-39021670 on Build GRCh38
rs987850836 Chr.22: 39021641-39021641 on Build GRCh38
rs953058600 Chr.22: 39021591-39021591 on Build GRCh38
rs937344241 Chr.22: 39021671-39021671 on Build GRCh38
rs373317174 Chr.22: 39021501-39021501 on Build GRCh38
rs762347432 Chr.22: 39021642-39021642 on Build GRCh38
rs376587988 Chr.22: 39021548-39021549 on Build GRCh38
rs370360323 Chr.22: 39021557-39021557 on Build GRCh38
rs1014726784 Chr.22: 39021560-39021560 on Build GRCh38
rs760983542 Chr.22: 39021520-39021520 on Build GRCh38
rs760959455 Chr.22: 39021530-39021530 on Build GRCh38
rs536287941 Chr.22: 39021666-39021666 on Build GRCh38
rs374897419 Chr.22: 39021541-39021541 on Build GRCh38
rs753610470 Chr.22: 39021563-39021564 on Build GRCh38
rs911668203 Chr.22: 39021452-39021452 on Build GRCh38
rs921581589 Chr.22: 39021437-39021437 on Build GRCh38
rs550876955 Chr.22: 39021632-39021632 on Build GRCh38
rs370204193 Chr.22: 39021537-39021537 on Build GRCh38
rs768851673 Chr.22: 39021552-39021552 on Build GRCh38
rs535434599 Chr.22: 39021569-39021569 on Build GRCh38
rs372799522 Chr.22: 39021630-39021630 on Build GRCh38
rs371316082 Chr.22: 39021443-39021443 on Build GRCh38
rs6001387 Chr.22: 39021506-39021506 on Build GRCh38
rs565089488 Chr.22: 39021558-39021558 on Build GRCh38
rs947622756 Chr.22: 39021522-39021522 on Build GRCh38
rs746181366 Chr.22: 39021570-39021570 on Build GRCh38
rs777063634 Chr.22: 39021584-39021584 on Build GRCh38
rs142926826 Chr.22: 39021579-39021579 on Build GRCh38
rs368214092 Chr.22: 39021572-39021572 on Build GRCh38
rs560480371 Chr.22: 39021544-39021544 on Build GRCh38
rs772188291 Chr.22: 39021585-39021585 on Build GRCh38
rs774099387 Chr.22: 39021476-39021476 on Build GRCh38
rs987572858 Chr.22: 39021612-39021612 on Build GRCh38
rs777250595 Chr.22: 39021561-39021561 on Build GRCh38
rs767701662 Chr.22: 39021502-39021502 on Build GRCh38
rs574805799 Chr.22: 39021531-39021531 on Build GRCh38
rs1053186746 Chr.22: 39021487-39021487 on Build GRCh38
rs911573339 Chr.22: 39021627-39021627 on Build GRCh38
rs5995657 Chr.22: 39021649-39021649 on Build GRCh38
rs199852673 Chr.22: 39021554-39021554 on Build GRCh38
rs750879061 Chr.22: 39021549-39021549 on Build GRCh38

View Full Product Details