Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65581861-65582134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VWA9
Entrez Gene ID
81556
Gene Name
von Willebrand factor A domain containing 9
Gene Aliases
C15orf44
UniGene
Hs.6686
Gene Chromosome Location
Chr.15: 65578757-65611289 on Build GRCh38
Transcripts
NM_001136043.2 NM_001207058.1 NM_001207059.1 NR_045104.1 NR_045105.2 NR_045106.2 NR_045901.1 XM_006720708.3 XM_006720709.3 XM_006720711.2 XM_011522101.2 XM_017022644.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4666932 c.1120_1121delCA p.Q374fs*4 Chr.15: 65581967-65581968 on Build GRCh38
964080 c.1262G>T p.R421I Chr.15: 65581997-65581997 on Build GRCh38
6370892 c.1080G>T p.Q360H Chr.15: 65582008-65582008 on Build GRCh38
4851890 c.1413G>A p.K471K Chr.15: 65581954-65581954 on Build GRCh38
6370891 c.1251G>T p.Q417H Chr.15: 65582008-65582008 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781635472 Chr.15: 65581970-65581971 on Build GRCh38
rs184385880 Chr.15: 65582083-65582083 on Build GRCh38
rs774082157 Chr.15: 65582064-65582064 on Build GRCh38
rs1029614076 Chr.15: 65581916-65581916 on Build GRCh38
rs71139418 Chr.15: 65582030-65582031 on Build GRCh38
rs778407091 Chr.15: 65581968-65581968 on Build GRCh38
rs754477514 Chr.15: 65582065-65582065 on Build GRCh38
rs398027699 Chr.15: 65582041-65582042 on Build GRCh38
rs370904636 Chr.15: 65582045-65582045 on Build GRCh38
rs776494570 Chr.15: 65582036-65582036 on Build GRCh38
rs1025663925 Chr.15: 65582097-65582097 on Build GRCh38
rs202079698 Chr.15: 65581929-65581929 on Build GRCh38
rs774613796 Chr.15: 65581990-65581990 on Build GRCh38
rs755255162 Chr.15: 65581966-65581967 on Build GRCh38
rs771264076 Chr.15: 65581971-65581971 on Build GRCh38
rs760741517 Chr.15: 65582027-65582027 on Build GRCh38
rs575165215 Chr.15: 65582031-65582031 on Build GRCh38
rs201463298 Chr.15: 65582042-65582042 on Build GRCh38
rs746484964 Chr.15: 65581952-65581952 on Build GRCh38
rs750161429 Chr.15: 65582044-65582044 on Build GRCh38
rs376183630 Chr.15: 65581983-65581983 on Build GRCh38
rs778577321 Chr.15: 65581911-65581911 on Build GRCh38
rs746055995 Chr.15: 65581992-65581992 on Build GRCh38
rs761686872 Chr.15: 65582041-65582041 on Build GRCh38
rs962489053 Chr.15: 65582079-65582079 on Build GRCh38
rs766122527 Chr.15: 65582052-65582052 on Build GRCh38
rs758008085 Chr.15: 65581918-65581918 on Build GRCh38
rs757061881 Chr.15: 65581910-65581910 on Build GRCh38
rs770579717 Chr.15: 65582056-65582057 on Build GRCh38
rs768345712 Chr.15: 65581965-65581965 on Build GRCh38
rs763989496 Chr.15: 65581904-65581904 on Build GRCh38
rs532153145 Chr.15: 65582043-65582043 on Build GRCh38
rs753839738 Chr.15: 65581907-65581907 on Build GRCh38
rs796162124 Chr.15: 65582031-65582032 on Build GRCh38
rs779900348 Chr.15: 65581951-65581951 on Build GRCh38
rs749768548 Chr.15: 65581970-65581970 on Build GRCh38
rs375724752 Chr.15: 65582062-65582062 on Build GRCh38
rs747233717 Chr.15: 65581930-65581931 on Build GRCh38
rs571113868 Chr.15: 65581913-65581913 on Build GRCh38
rs544146113 Chr.15: 65582031-65582032 on Build GRCh38
rs371096322 Chr.15: 65582022-65582022 on Build GRCh38
rs764216824 Chr.15: 65582029-65582029 on Build GRCh38

View Full Product Details