Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 35312271-35312542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEF6
Entrez Gene ID
50619
Gene Name
DEF6, guanine nucleotide exchange factor
Gene Aliases
IBP, SLAT, SWAP70L
UniGene
Hs.15476
Gene Chromosome Location
Chr.6: 35297818-35321771 on Build GRCh38
Transcripts
NM_022047.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7364046 c.569C>T p.S190L Chr.6: 35312447-35312447 on Build GRCh38
364699 c.622G>A p.E208K Chr.6: 35312500-35312500 on Build GRCh38
1546477 c.595C>T p.R199W Chr.6: 35312473-35312473 on Build GRCh38
6547193 c.585G>T p.R195R Chr.6: 35312463-35312463 on Build GRCh38
6106773 c.589G>T p.V197L Chr.6: 35312467-35312467 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777889512 Chr.6: 35312384-35312384 on Build GRCh38
rs773044621 Chr.6: 35312452-35312452 on Build GRCh38
rs752905282 Chr.6: 35312466-35312466 on Build GRCh38
rs775030898 Chr.6: 35312292-35312292 on Build GRCh38
rs771987663 Chr.6: 35312448-35312448 on Build GRCh38
rs543938116 Chr.6: 35312461-35312461 on Build GRCh38
rs941634967 Chr.6: 35312451-35312451 on Build GRCh38
rs35395160 Chr.6: 35312406-35312406 on Build GRCh38
rs369847754 Chr.6: 35312339-35312339 on Build GRCh38
rs748995204 Chr.6: 35312429-35312429 on Build GRCh38
rs200254432 Chr.6: 35312467-35312467 on Build GRCh38
rs370895388 Chr.6: 35312298-35312298 on Build GRCh38
rs761895973 Chr.6: 35312332-35312332 on Build GRCh38
rs779727492 Chr.6: 35312419-35312419 on Build GRCh38
rs563519848 Chr.6: 35312499-35312499 on Build GRCh38
rs766609884 Chr.6: 35312406-35312407 on Build GRCh38
rs760399698 Chr.6: 35312354-35312354 on Build GRCh38
rs148839014 Chr.6: 35312439-35312439 on Build GRCh38
rs146706360 Chr.6: 35312430-35312430 on Build GRCh38
rs766125119 Chr.6: 35312462-35312462 on Build GRCh38
rs539849075 Chr.6: 35312408-35312408 on Build GRCh38
rs770090116 Chr.6: 35312412-35312412 on Build GRCh38
rs750935944 Chr.6: 35312496-35312496 on Build GRCh38
rs750165093 Chr.6: 35312341-35312341 on Build GRCh38
rs539688189 Chr.6: 35312325-35312325 on Build GRCh38
rs773902946 Chr.6: 35312375-35312376 on Build GRCh38
rs780620376 Chr.6: 35312500-35312500 on Build GRCh38
rs142511424 Chr.6: 35312475-35312475 on Build GRCh38
rs998640078 Chr.6: 35312493-35312493 on Build GRCh38
rs761408118 Chr.6: 35312447-35312447 on Build GRCh38
rs1023251355 Chr.6: 35312330-35312330 on Build GRCh38
rs1041454042 Chr.6: 35312474-35312474 on Build GRCh38
rs768325136 Chr.6: 35312308-35312308 on Build GRCh38
rs765449019 Chr.6: 35312355-35312355 on Build GRCh38
rs770896714 Chr.6: 35312481-35312481 on Build GRCh38
rs752946127 Chr.6: 35312364-35312364 on Build GRCh38
rs764306600 Chr.6: 35312473-35312473 on Build GRCh38
rs557782882 Chr.6: 35312376-35312376 on Build GRCh38
rs746197755 Chr.6: 35312409-35312409 on Build GRCh38
rs1043201299 Chr.6: 35312453-35312453 on Build GRCh38
rs768124826 Chr.6: 35312345-35312346 on Build GRCh38
rs377311393 Chr.6: 35312482-35312482 on Build GRCh38
rs144395191 Chr.6: 35312407-35312407 on Build GRCh38
rs982188920 Chr.6: 35312347-35312347 on Build GRCh38
rs761086759 Chr.6: 35312376-35312384 on Build GRCh38

View Full Product Details