Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 238128750-238128975 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESPNL
Entrez Gene ID
339768
Gene Name
espin-like
Gene Aliases
-
UniGene
Hs.127724
Gene Chromosome Location
Chr.2: 238100310-238133287 on Build GRCh38
Transcripts
NM_001308370.1 NM_194312.3 XM_011511087.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5035409 c.1393G>A p.E465K Chr.2: 238128884-238128884 on Build GRCh38
4624688 c.1275T>C p.D425D Chr.2: 238128766-238128766 on Build GRCh38
1165327 c.1314G>A p.T438T Chr.2: 238128805-238128805 on Build GRCh38
7414492 c.1303C>A p.L435M Chr.2: 238128794-238128794 on Build GRCh38
4966304 c.1324C>T p.R442C Chr.2: 238128815-238128815 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1031013473 Chr.2: 238128899-238128899 on Build GRCh38
rs745776811 Chr.2: 238128808-238128809 on Build GRCh38
rs926880383 Chr.2: 238128907-238128907 on Build GRCh38
rs771283994 Chr.2: 238128851-238128851 on Build GRCh38
rs61571075 Chr.2: 238128781-238128781 on Build GRCh38
rs988275970 Chr.2: 238128882-238128882 on Build GRCh38
rs540187196 Chr.2: 238128843-238128843 on Build GRCh38
rs1014733271 Chr.2: 238128780-238128780 on Build GRCh38
rs990088528 Chr.2: 238128910-238128910 on Build GRCh38
rs749131183 Chr.2: 238128910-238128910 on Build GRCh38
rs1014048204 Chr.2: 238128928-238128928 on Build GRCh38
rs374688238 Chr.2: 238128791-238128791 on Build GRCh38
rs367647458 Chr.2: 238128818-238128818 on Build GRCh38
rs986065286 Chr.2: 238128831-238128831 on Build GRCh38
rs1044194444 Chr.2: 238128807-238128807 on Build GRCh38
rs530876530 Chr.2: 238128943-238128943 on Build GRCh38
rs371572172 Chr.2: 238128788-238128788 on Build GRCh38
rs768608693 Chr.2: 238128896-238128896 on Build GRCh38
rs1055431212 Chr.2: 238128923-238128923 on Build GRCh38
rs767293678 Chr.2: 238128944-238128944 on Build GRCh38
rs764172391 Chr.2: 238128787-238128787 on Build GRCh38
rs745929145 Chr.2: 238128863-238128863 on Build GRCh38
rs749122984 Chr.2: 238128815-238128815 on Build GRCh38
rs772847700 Chr.2: 238128946-238128946 on Build GRCh38
rs1020156242 Chr.2: 238128878-238128878 on Build GRCh38
rs914089002 Chr.2: 238128890-238128890 on Build GRCh38
rs1014101896 Chr.2: 238128778-238128778 on Build GRCh38
rs749848965 Chr.2: 238128792-238128792 on Build GRCh38
rs761619561 Chr.2: 238128790-238128790 on Build GRCh38
rs889982364 Chr.2: 238128875-238128875 on Build GRCh38
rs545228236 Chr.2: 238128797-238128797 on Build GRCh38
rs955916165 Chr.2: 238128909-238128909 on Build GRCh38
rs758740622 Chr.2: 238128825-238128825 on Build GRCh38
rs527619102 Chr.2: 238128932-238128933 on Build GRCh38
rs966060820 Chr.2: 238128873-238128873 on Build GRCh38
rs561531418 Chr.2: 238128858-238128858 on Build GRCh38
rs901260203 Chr.2: 238128879-238128879 on Build GRCh38
rs147504541 Chr.2: 238128927-238128927 on Build GRCh38
rs1021453796 Chr.2: 238128917-238128917 on Build GRCh38
rs911995537 Chr.2: 238128834-238128834 on Build GRCh38
rs775828964 Chr.2: 238128905-238128905 on Build GRCh38
rs918283670 Chr.2: 238128952-238128952 on Build GRCh38
rs774323350 Chr.2: 238128921-238128921 on Build GRCh38
rs867316250 Chr.2: 238128786-238128786 on Build GRCh38
rs914422355 Chr.2: 238128768-238128768 on Build GRCh38
rs770109267 Chr.2: 238128900-238128900 on Build GRCh38
rs1006922064 Chr.2: 238128876-238128876 on Build GRCh38
rs200132075 Chr.2: 238128820-238128820 on Build GRCh38
rs937001731 Chr.2: 238128922-238128922 on Build GRCh38
rs779052998 Chr.2: 238128779-238128781 on Build GRCh38
rs768607011 Chr.2: 238128911-238128911 on Build GRCh38
rs572667961 Chr.2: 238128827-238128827 on Build GRCh38
rs1036699908 Chr.2: 238128842-238128842 on Build GRCh38
rs550687242 Chr.2: 238128929-238128929 on Build GRCh38
rs766704104 Chr.2: 238128935-238128935 on Build GRCh38
rs1048527234 Chr.2: 238128860-238128860 on Build GRCh38
rs779679192 Chr.2: 238128800-238128800 on Build GRCh38

View Full Product Details