Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 112168544-112168811 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPB41L4A
Entrez Gene ID
64097
Gene Name
erythrocyte membrane protein band 4.1 like 4A
Gene Aliases
EPB41L4, NBL4
UniGene
Hs.584954
Gene Chromosome Location
Chr.5: 112141829-112419490 on Build GRCh38
Transcripts
NM_022140.3 XM_011543530.1 XM_011543531.2 XM_011543532.1 XM_011543533.1 XM_017009689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6314898 c.1886C>A p.T629N Chr.5: 112168785-112168785 on Build GRCh38
6214843 c.1913G>C p.G638A Chr.5: 112168758-112168758 on Build GRCh38
1432114 c.1896G>A p.S632S Chr.5: 112168775-112168775 on Build GRCh38
6314899 c.767C>A p.T256N Chr.5: 112168785-112168785 on Build GRCh38
6214844 c.794G>C p.G265A Chr.5: 112168758-112168758 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768025406 Chr.5: 112168756-112168756 on Build GRCh38
rs368861569 Chr.5: 112168776-112168776 on Build GRCh38
rs748912347 Chr.5: 112168770-112168770 on Build GRCh38
rs750945497 Chr.5: 112168761-112168761 on Build GRCh38
rs944927008 Chr.5: 112168617-112168617 on Build GRCh38
rs7703522 Chr.5: 112168782-112168782 on Build GRCh38
rs922120450 Chr.5: 112168646-112168646 on Build GRCh38
rs375125928 Chr.5: 112168709-112168709 on Build GRCh38
rs747527138 Chr.5: 112168597-112168597 on Build GRCh38
rs17319543 Chr.5: 112168706-112168706 on Build GRCh38
rs1011239274 Chr.5: 112168661-112168661 on Build GRCh38
rs766288959 Chr.5: 112168725-112168725 on Build GRCh38
rs867657799 Chr.5: 112168659-112168659 on Build GRCh38
rs757303695 Chr.5: 112168754-112168754 on Build GRCh38
rs953064855 Chr.5: 112168574-112168575 on Build GRCh38
rs966328572 Chr.5: 112168596-112168596 on Build GRCh38
rs763301958 Chr.5: 112168741-112168741 on Build GRCh38
rs538729529 Chr.5: 112168663-112168663 on Build GRCh38
rs749000368 Chr.5: 112168690-112168690 on Build GRCh38
rs935085472 Chr.5: 112168614-112168614 on Build GRCh38
rs947732956 Chr.5: 112168606-112168606 on Build GRCh38
rs918777741 Chr.5: 112168572-112168572 on Build GRCh38
rs1050399573 Chr.5: 112168676-112168676 on Build GRCh38
rs775841254 Chr.5: 112168740-112168740 on Build GRCh38
rs886443621 Chr.5: 112168724-112168724 on Build GRCh38
rs771478027 Chr.5: 112168723-112168724 on Build GRCh38
rs749864627 Chr.5: 112168721-112168722 on Build GRCh38
rs756624587 Chr.5: 112168694-112168695 on Build GRCh38
rs180931108 Chr.5: 112168747-112168747 on Build GRCh38
rs768050533 Chr.5: 112168702-112168702 on Build GRCh38
rs912118080 Chr.5: 112168608-112168608 on Build GRCh38
rs975340167 Chr.5: 112168640-112168640 on Build GRCh38
rs766163075 Chr.5: 112168783-112168783 on Build GRCh38
rs544419945 Chr.5: 112168790-112168790 on Build GRCh38
rs778452899 Chr.5: 112168775-112168775 on Build GRCh38
rs752316139 Chr.5: 112168713-112168715 on Build GRCh38
rs1019593421 Chr.5: 112168732-112168732 on Build GRCh38
rs868429825 Chr.5: 112168759-112168759 on Build GRCh38
rs756460326 Chr.5: 112168765-112168765 on Build GRCh38
rs927659727 Chr.5: 112168611-112168611 on Build GRCh38
rs780468584 Chr.5: 112168769-112168769 on Build GRCh38
rs189409879 Chr.5: 112168746-112168746 on Build GRCh38
rs571434821 Chr.5: 112168588-112168588 on Build GRCh38
rs760481013 Chr.5: 112168715-112168715 on Build GRCh38
rs1052668635 Chr.5: 112168628-112168628 on Build GRCh38
rs893667621 Chr.5: 112168644-112168644 on Build GRCh38
rs572106322 Chr.5: 112168712-112168712 on Build GRCh38
rs879217234 Chr.5: 112168748-112168748 on Build GRCh38
rs773316110 Chr.5: 112168714-112168714 on Build GRCh38
rs772818883 Chr.5: 112168724-112168727 on Build GRCh38
rs754709341 Chr.5: 112168771-112168771 on Build GRCh38
rs773955133 Chr.5: 112168705-112168705 on Build GRCh38

View Full Product Details