Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 18510663-18510930 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKL5
Entrez Gene ID
6792
Gene Name
cyclin dependent kinase like 5
Gene Aliases
CFAP247, EIEE2, ISSX, STK9
UniGene
Hs.659851
Gene Chromosome Location
Chr.X: 18425605-18653629 on Build GRCh38
Transcripts
NM_001037343.1 NM_001323289.1 NM_003159.2

SNP Detail

SNP ID Chromosome Location
rs753796357 Chr.X: 18510734-18510734 on Build GRCh38
rs368320902 Chr.X: 18510771-18510771 on Build GRCh38
rs769668864 Chr.X: 18510790-18510790 on Build GRCh38
rs587783130 Chr.X: 18510828-18510828 on Build GRCh38
rs891192393 Chr.X: 18510845-18510845 on Build GRCh38
rs752513906 Chr.X: 18510791-18510791 on Build GRCh38
rs587783131 Chr.X: 18510859-18510859 on Build GRCh38
rs371743051 Chr.X: 18510782-18510782 on Build GRCh38
rs786204991 Chr.X: 18510846-18510846 on Build GRCh38
rs768612597 Chr.X: 18510900-18510900 on Build GRCh38
rs777490768 Chr.X: 18510816-18510816 on Build GRCh38
rs532222548 Chr.X: 18510807-18510807 on Build GRCh38
rs529850269 Chr.X: 18510706-18510706 on Build GRCh38
rs746294282 Chr.X: 18510731-18510731 on Build GRCh38
rs756095341 Chr.X: 18510792-18510792 on Build GRCh38
rs970189664 Chr.X: 18510697-18510697 on Build GRCh38
rs267608422 Chr.X: 18510883-18510883 on Build GRCh38
rs267608420 Chr.X: 18510820-18510821 on Build GRCh38
rs140332992 Chr.X: 18510848-18510848 on Build GRCh38
rs767125302 Chr.X: 18510787-18510787 on Build GRCh38
rs757081261 Chr.X: 18510877-18510877 on Build GRCh38
rs369016064 Chr.X: 18510904-18510904 on Build GRCh38
rs776029783 Chr.X: 18510803-18510803 on Build GRCh38
rs754436143 Chr.X: 18510786-18510786 on Build GRCh38
rs267608421 Chr.X: 18510855-18510855 on Build GRCh38
rs137874941 Chr.X: 18510888-18510888 on Build GRCh38
rs376559686 Chr.X: 18510760-18510760 on Build GRCh38
rs907569051 Chr.X: 18510774-18510774 on Build GRCh38

View Full Product Details