Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26440231-26440458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRC1
Entrez Gene ID
92749
Gene Name
dynein regulatory complex subunit 1
Gene Aliases
C2orf39, CCDC164, CILD21
UniGene
Hs.393714
Gene Chromosome Location
Chr.2: 26401912-26456713 on Build GRCh38
Transcripts
NM_145038.4 XM_005264637.3 XM_005264638.3 XM_017005271.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6649243 c.951G>A p.E317E Chr.2: 26440440-26440440 on Build GRCh38
6649241 c.949G>A p.E317K Chr.2: 26440438-26440438 on Build GRCh38
5917557 c.937C>T p.Q313* Chr.2: 26440426-26440426 on Build GRCh38
1019357 c.942G>T p.E314D Chr.2: 26440431-26440431 on Build GRCh38
5862292 c.892C>T p.L298F Chr.2: 26440381-26440381 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761334645 Chr.2: 26440355-26440355 on Build GRCh38
rs139352961 Chr.2: 26440398-26440398 on Build GRCh38
rs773063930 Chr.2: 26440363-26440363 on Build GRCh38
rs149216279 Chr.2: 26440277-26440277 on Build GRCh38
rs755289126 Chr.2: 26440340-26440347 on Build GRCh38
rs757321423 Chr.2: 26440401-26440401 on Build GRCh38
rs781514755 Chr.2: 26440342-26440349 on Build GRCh38
rs538651314 Chr.2: 26440343-26440343 on Build GRCh38
rs59636742 Chr.2: 26440345-26440346 on Build GRCh38
rs774671488 Chr.2: 26440363-26440363 on Build GRCh38
rs751954182 Chr.2: 26440338-26440347 on Build GRCh38
rs749272904 Chr.2: 26440346-26440349 on Build GRCh38
rs374305580 Chr.2: 26440315-26440315 on Build GRCh38
rs748730164 Chr.2: 26440344-26440347 on Build GRCh38
rs769322821 Chr.2: 26440357-26440357 on Build GRCh38
rs56218945 Chr.2: 26440347-26440347 on Build GRCh38
rs775977815 Chr.2: 26440353-26440353 on Build GRCh38
rs144917673 Chr.2: 26440276-26440276 on Build GRCh38
rs546940443 Chr.2: 26440301-26440301 on Build GRCh38
rs200704313 Chr.2: 26440319-26440319 on Build GRCh38
rs762889685 Chr.2: 26440369-26440369 on Build GRCh38
rs13421454 Chr.2: 26440345-26440345 on Build GRCh38
rs796367128 Chr.2: 26440345-26440346 on Build GRCh38
rs746818380 Chr.2: 26440362-26440363 on Build GRCh38
rs779140891 Chr.2: 26440407-26440407 on Build GRCh38
rs780878055 Chr.2: 26440329-26440330 on Build GRCh38
rs58723978 Chr.2: 26440346-26440347 on Build GRCh38
rs370717886 Chr.2: 26440387-26440387 on Build GRCh38
rs72217381 Chr.2: 26440320-26440321 on Build GRCh38
rs373874150 Chr.2: 26440389-26440389 on Build GRCh38
rs746409714 Chr.2: 26440339-26440339 on Build GRCh38
rs70950194 Chr.2: 26440361-26440362 on Build GRCh38
rs138497746 Chr.2: 26440319-26440320 on Build GRCh38
rs1002193111 Chr.2: 26440323-26440323 on Build GRCh38
rs201498921 Chr.2: 26440349-26440349 on Build GRCh38
rs764426254 Chr.2: 26440391-26440391 on Build GRCh38
rs766148507 Chr.2: 26440371-26440371 on Build GRCh38
rs373963153 Chr.2: 26440351-26440351 on Build GRCh38

View Full Product Details