Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119097157-119097406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPAGT1
Entrez Gene ID
1798
Gene Name
dolichyl-phosphate N-acetylglucosaminephosphotransferase 1
Gene Aliases
ALG7, CDG-Ij, CDG1J, CMS13, CMSTA2, D11S366, DGPT, DPAGT, DPAGT2, G1PT, GPT, UAGT, UGAT
UniGene
Hs.524081
Gene Chromosome Location
Chr.11: 119096503-119102414 on Build GRCh38
Transcripts
NM_001382.3 XM_005271422.2 XM_011542648.1 XM_017017293.1 XM_017017294.1 XM_017017295.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5573430 c.685-2A>T p.? Chr.11: 119097299-119097299 on Build GRCh38
6500829 c.697C>T p.L233F Chr.11: 119097285-119097285 on Build GRCh38
5201157 c.798delC p.I267fs*15 Chr.11: 119097184-119097184 on Build GRCh38
5573429 c.1006-2A>T p.? Chr.11: 119097299-119097299 on Build GRCh38
3375680 c.1049C>T p.T350I Chr.11: 119097254-119097254 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748295799 Chr.11: 119097332-119097332 on Build GRCh38
rs780550367 Chr.11: 119097313-119097313 on Build GRCh38
rs376996734 Chr.11: 119097285-119097285 on Build GRCh38
rs980077282 Chr.11: 119097204-119097204 on Build GRCh38
rs574176195 Chr.11: 119097323-119097323 on Build GRCh38
rs543421224 Chr.11: 119097260-119097260 on Build GRCh38
rs917922996 Chr.11: 119097355-119097355 on Build GRCh38
rs770121489 Chr.11: 119097338-119097338 on Build GRCh38
rs77836371 Chr.11: 119097300-119097300 on Build GRCh38
rs758885855 Chr.11: 119097312-119097312 on Build GRCh38
rs778250734 Chr.11: 119097304-119097304 on Build GRCh38
rs141849365 Chr.11: 119097374-119097374 on Build GRCh38
rs555841797 Chr.11: 119097279-119097279 on Build GRCh38
rs745447691 Chr.11: 119097311-119097311 on Build GRCh38
rs943371372 Chr.11: 119097339-119097339 on Build GRCh38
rs776863215 Chr.11: 119097324-119097324 on Build GRCh38
rs777530944 Chr.11: 119097277-119097277 on Build GRCh38
rs773152872 Chr.11: 119097340-119097340 on Build GRCh38
rs752484662 Chr.11: 119097254-119097254 on Build GRCh38
rs139295058 Chr.11: 119097192-119097192 on Build GRCh38

View Full Product Details