Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 30379389-30379656 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPD
Entrez Gene ID
1362
Gene Name
carboxypeptidase D
Gene Aliases
GP180
UniGene
Hs.446079
Gene Chromosome Location
Chr.17: 30378924-30469657 on Build GRCh38
Transcripts
NM_001304.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4613034 c.558_559insT p.E188fs*24 Chr.17: 30379538-30379539 on Build GRCh38
4151572 c.410A>G p.N137S Chr.17: 30379390-30379390 on Build GRCh38
6410754 c.543C>A p.S181R Chr.17: 30379523-30379523 on Build GRCh38
6016557 c.579C>T p.G193G Chr.17: 30379559-30379559 on Build GRCh38
5601301 c.514A>T p.N172Y Chr.17: 30379494-30379494 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549565879 Chr.17: 30379571-30379571 on Build GRCh38
rs769306492 Chr.17: 30379469-30379469 on Build GRCh38
rs748073994 Chr.17: 30379574-30379574 on Build GRCh38
rs747304806 Chr.17: 30379589-30379589 on Build GRCh38
rs563958469 Chr.17: 30379471-30379471 on Build GRCh38
rs759561934 Chr.17: 30379609-30379609 on Build GRCh38
rs779470503 Chr.17: 30379475-30379475 on Build GRCh38
rs748726965 Chr.17: 30379556-30379556 on Build GRCh38
rs760926393 Chr.17: 30379502-30379502 on Build GRCh38
rs771953389 Chr.17: 30379497-30379497 on Build GRCh38
rs775875206 Chr.17: 30379612-30379612 on Build GRCh38
rs752978220 Chr.17: 30379454-30379454 on Build GRCh38
rs370241648 Chr.17: 30379501-30379501 on Build GRCh38
rs538272933 Chr.17: 30379597-30379614 on Build GRCh38
rs922272566 Chr.17: 30379415-30379415 on Build GRCh38
rs531434261 Chr.17: 30379551-30379551 on Build GRCh38
rs373926366 Chr.17: 30379481-30379481 on Build GRCh38
rs754984161 Chr.17: 30379568-30379568 on Build GRCh38
rs373064492 Chr.17: 30379440-30379440 on Build GRCh38
rs765628135 Chr.17: 30379527-30379527 on Build GRCh38
rs774252098 Chr.17: 30379482-30379482 on Build GRCh38
rs182416584 Chr.17: 30379441-30379441 on Build GRCh38
rs762154184 Chr.17: 30379428-30379428 on Build GRCh38
rs761482605 Chr.17: 30379493-30379493 on Build GRCh38
rs751905527 Chr.17: 30379628-30379628 on Build GRCh38
rs765308271 Chr.17: 30379610-30379610 on Build GRCh38
rs910769351 Chr.17: 30379459-30379459 on Build GRCh38
rs755815841 Chr.17: 30379636-30379636 on Build GRCh38
rs753205118 Chr.17: 30379542-30379542 on Build GRCh38
rs755375779 Chr.17: 30379449-30379449 on Build GRCh38
rs756971273 Chr.17: 30379461-30379461 on Build GRCh38
rs962459723 Chr.17: 30379611-30379611 on Build GRCh38
rs960942003 Chr.17: 30379607-30379607 on Build GRCh38
rs750079998 Chr.17: 30379549-30379549 on Build GRCh38
rs766574228 Chr.17: 30379508-30379508 on Build GRCh38
rs906548606 Chr.17: 30379524-30379524 on Build GRCh38
rs764508943 Chr.17: 30379622-30379622 on Build GRCh38
rs753860996 Chr.17: 30379510-30379510 on Build GRCh38
rs754386584 Chr.17: 30379448-30379448 on Build GRCh38
rs745473994 Chr.17: 30379467-30379467 on Build GRCh38
rs779390393 Chr.17: 30379450-30379450 on Build GRCh38
rs771254289 Chr.17: 30379591-30379591 on Build GRCh38
rs750571315 Chr.17: 30379431-30379431 on Build GRCh38
rs1016751702 Chr.17: 30379592-30379592 on Build GRCh38
rs772010503 Chr.17: 30379575-30379575 on Build GRCh38
rs767826268 Chr.17: 30379430-30379430 on Build GRCh38
rs780850763 Chr.17: 30379466-30379466 on Build GRCh38
rs776809492 Chr.17: 30379593-30379593 on Build GRCh38
rs749312900 Chr.17: 30379477-30379477 on Build GRCh38
rs754086114 Chr.17: 30379458-30379458 on Build GRCh38
rs140283576 Chr.17: 30379514-30379514 on Build GRCh38
rs990982154 Chr.17: 30379608-30379608 on Build GRCh38
rs773078547 Chr.17: 30379585-30379585 on Build GRCh38
rs755758637 Chr.17: 30379550-30379550 on Build GRCh38
rs758779593 Chr.17: 30379547-30379547 on Build GRCh38

View Full Product Details