Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 207570224-207570449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREB1
Entrez Gene ID
1385
Gene Name
cAMP responsive element binding protein 1
Gene Aliases
CREB, CREB-1
UniGene
Hs.516646
Gene Chromosome Location
Chr.2: 207529892-207605989 on Build GRCh38
Transcripts
NM_001320793.1 NM_004379.4 NM_134442.4 NR_135473.1 XM_011510645.1 XM_011510646.2 XM_011510647.2 XM_011510648.2 XM_011510649.2 XM_011510650.2 XM_011510651.2 XM_017003399.1 XM_017003400.1 XM_017003401.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6292769 c.547+3G>T p.? Chr.2: 207570324-207570324 on Build GRCh38
4829726 c.454G>C p.E152Q Chr.2: 207570228-207570228 on Build GRCh38
5481684 c.506C>T p.T169M Chr.2: 207570280-207570280 on Build GRCh38
7328181 c.547+1G>T p.? Chr.2: 207570322-207570322 on Build GRCh38
3838426 c.498C>T p.T166T Chr.2: 207570272-207570272 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752980714 Chr.2: 207570363-207570363 on Build GRCh38
rs761154605 Chr.2: 207570334-207570334 on Build GRCh38
rs552888247 Chr.2: 207570372-207570372 on Build GRCh38
rs942074081 Chr.2: 207570253-207570253 on Build GRCh38
rs779699990 Chr.2: 207570260-207570260 on Build GRCh38
rs201006565 Chr.2: 207570388-207570388 on Build GRCh38
rs201642429 Chr.2: 207570281-207570281 on Build GRCh38
rs113872103 Chr.2: 207570417-207570417 on Build GRCh38
rs896948175 Chr.2: 207570329-207570329 on Build GRCh38
rs765332411 Chr.2: 207570354-207570354 on Build GRCh38
rs748868942 Chr.2: 207570270-207570270 on Build GRCh38
rs767690757 Chr.2: 207570359-207570359 on Build GRCh38
rs766917972 Chr.2: 207570343-207570343 on Build GRCh38
rs769918733 Chr.2: 207570258-207570258 on Build GRCh38
rs995126472 Chr.2: 207570330-207570330 on Build GRCh38
rs773628752 Chr.2: 207570333-207570333 on Build GRCh38
rs752889040 Chr.2: 207570373-207570373 on Build GRCh38
rs762025997 Chr.2: 207570282-207570282 on Build GRCh38
rs759695908 Chr.2: 207570350-207570350 on Build GRCh38
rs759621519 Chr.2: 207570351-207570364 on Build GRCh38
rs899292076 Chr.2: 207570264-207570264 on Build GRCh38
rs772427535 Chr.2: 207570302-207570302 on Build GRCh38
rs757047432 Chr.2: 207570365-207570365 on Build GRCh38
rs753834321 Chr.2: 207570346-207570346 on Build GRCh38
rs192210277 Chr.2: 207570389-207570389 on Build GRCh38
rs1017878574 Chr.2: 207570405-207570405 on Build GRCh38
rs950458883 Chr.2: 207570402-207570402 on Build GRCh38
rs768152463 Chr.2: 207570272-207570272 on Build GRCh38
rs967788255 Chr.2: 207570419-207570419 on Build GRCh38

View Full Product Details