Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43974930-43975179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V0B
Entrez Gene ID
533
Gene Name
ATPase H+ transporting V0 subunit b
Gene Aliases
ATP6F, HATPL, VMA16
UniGene
Hs.596514
Gene Chromosome Location
Chr.1: 43974648-43978300 on Build GRCh38
Transcripts
NM_001039457.2 NM_001294333.1 NM_004047.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7313146 c.6G>C p.T2T Chr.1: 43975046-43975046 on Build GRCh38
3751232 c.57G>C p.A19A Chr.1: 43975097-43975097 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs895404941 Chr.1: 43974963-43974963 on Build GRCh38
rs1045119218 Chr.1: 43975163-43975163 on Build GRCh38
rs898468575 Chr.1: 43974989-43974989 on Build GRCh38
rs1019159397 Chr.1: 43975070-43975070 on Build GRCh38
rs961821749 Chr.1: 43975020-43975020 on Build GRCh38
rs923683979 Chr.1: 43975162-43975162 on Build GRCh38
rs762606732 Chr.1: 43975098-43975098 on Build GRCh38
rs935358675 Chr.1: 43974974-43974974 on Build GRCh38
rs547891031 Chr.1: 43974961-43974961 on Build GRCh38
rs11549137 Chr.1: 43975038-43975038 on Build GRCh38
rs769570289 Chr.1: 43975094-43975094 on Build GRCh38
rs979345872 Chr.1: 43974954-43974954 on Build GRCh38
rs977870549 Chr.1: 43975151-43975151 on Build GRCh38
rs908971622 Chr.1: 43974985-43974985 on Build GRCh38
rs771994652 Chr.1: 43975013-43975013 on Build GRCh38
rs72890650 Chr.1: 43974970-43974970 on Build GRCh38
rs1021984547 Chr.1: 43975118-43975118 on Build GRCh38
rs1025674085 Chr.1: 43974992-43974992 on Build GRCh38
rs1038387915 Chr.1: 43974978-43974978 on Build GRCh38
rs1008685570 Chr.1: 43975032-43975032 on Build GRCh38
rs1056773823 Chr.1: 43974962-43974962 on Build GRCh38
rs946530073 Chr.1: 43974950-43974950 on Build GRCh38
rs780026637 Chr.1: 43975049-43975049 on Build GRCh38
rs776524790 Chr.1: 43975077-43975077 on Build GRCh38
rs147249398 Chr.1: 43975020-43975021 on Build GRCh38
rs768112834 Chr.1: 43975106-43975106 on Build GRCh38
rs549721163 Chr.1: 43975045-43975045 on Build GRCh38
rs761273764 Chr.1: 43975147-43975147 on Build GRCh38
rs866801704 Chr.1: 43975141-43975141 on Build GRCh38
rs886311215 Chr.1: 43975005-43975005 on Build GRCh38
rs569887950 Chr.1: 43975073-43975073 on Build GRCh38
rs994239152 Chr.1: 43974990-43974990 on Build GRCh38
rs746800331 Chr.1: 43975054-43975054 on Build GRCh38
rs2286241 Chr.1: 43975097-43975097 on Build GRCh38
rs376125990 Chr.1: 43975144-43975144 on Build GRCh38
rs529893444 Chr.1: 43974981-43974981 on Build GRCh38

View Full Product Details