Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 56452127-56452392 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASS4
Entrez Gene ID
57091
Gene Name
Cas scaffolding protein family member 4
Gene Aliases
C20orf32, CAS4, HEFL, HEPL
UniGene
Hs.473144
Gene Chromosome Location
Chr.20: 56411550-56460240 on Build GRCh38
Transcripts
NM_001164114.1 NM_001164115.1 NM_001164116.1 NM_020356.3 XM_006723831.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6558906 c.1087G>T p.G363W Chr.20: 56452263-56452263 on Build GRCh38
1495136 c.1149G>A p.W383* Chr.20: 56452325-56452325 on Build GRCh38
5892429 c.1012C>T p.P338S Chr.20: 56452188-56452188 on Build GRCh38
6525971 c.1177C>A p.P393T Chr.20: 56452353-56452353 on Build GRCh38
3548039 c.1016G>A p.R339Q Chr.20: 56452192-56452192 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745844917 Chr.20: 56452193-56452193 on Build GRCh38
rs751309869 Chr.20: 56452286-56452286 on Build GRCh38
rs746817385 Chr.20: 56452200-56452200 on Build GRCh38
rs756187603 Chr.20: 56452248-56452248 on Build GRCh38
rs753845628 Chr.20: 56452343-56452343 on Build GRCh38
rs768228596 Chr.20: 56452210-56452210 on Build GRCh38
rs895670548 Chr.20: 56452325-56452325 on Build GRCh38
rs138854202 Chr.20: 56452247-56452247 on Build GRCh38
rs779877573 Chr.20: 56452196-56452196 on Build GRCh38
rs758270758 Chr.20: 56452260-56452260 on Build GRCh38
rs747825213 Chr.20: 56452305-56452305 on Build GRCh38
rs912723811 Chr.20: 56452242-56452242 on Build GRCh38
rs372419278 Chr.20: 56452320-56452320 on Build GRCh38
rs752780274 Chr.20: 56452180-56452180 on Build GRCh38
rs780870845 Chr.20: 56452303-56452303 on Build GRCh38
rs754781588 Chr.20: 56452289-56452289 on Build GRCh38
rs764387425 Chr.20: 56452340-56452340 on Build GRCh38
rs930994900 Chr.20: 56452262-56452262 on Build GRCh38
rs371177644 Chr.20: 56452244-56452244 on Build GRCh38
rs376949340 Chr.20: 56452355-56452355 on Build GRCh38
rs761910658 Chr.20: 56452349-56452349 on Build GRCh38
rs772583331 Chr.20: 56452368-56452368 on Build GRCh38
rs771526401 Chr.20: 56452312-56452312 on Build GRCh38
rs147388572 Chr.20: 56452361-56452361 on Build GRCh38
rs148219392 Chr.20: 56452259-56452259 on Build GRCh38
rs767728589 Chr.20: 56452246-56452246 on Build GRCh38
rs1057376930 Chr.20: 56452261-56452261 on Build GRCh38
rs1022868200 Chr.20: 56452347-56452347 on Build GRCh38
rs377289897 Chr.20: 56452233-56452233 on Build GRCh38
rs764146773 Chr.20: 56452255-56452255 on Build GRCh38
rs942097864 Chr.20: 56452230-56452230 on Build GRCh38
rs773768954 Chr.20: 56452232-56452232 on Build GRCh38
rs775776673 Chr.20: 56452336-56452336 on Build GRCh38
rs267606009 Chr.20: 56452337-56452337 on Build GRCh38
rs145924473 Chr.20: 56452229-56452229 on Build GRCh38
rs969386665 Chr.20: 56452175-56452175 on Build GRCh38
rs754801286 Chr.20: 56452360-56452360 on Build GRCh38
rs772361440 Chr.20: 56452335-56452335 on Build GRCh38
rs755196041 Chr.20: 56452168-56452168 on Build GRCh38
rs759955848 Chr.20: 56452331-56452331 on Build GRCh38
rs767006633 Chr.20: 56452238-56452238 on Build GRCh38
rs748813329 Chr.20: 56452307-56452307 on Build GRCh38
rs116883472 Chr.20: 56452211-56452211 on Build GRCh38
rs769333762 Chr.20: 56452306-56452306 on Build GRCh38
rs144494485 Chr.20: 56452194-56452194 on Build GRCh38
rs774792011 Chr.20: 56452243-56452243 on Build GRCh38
rs149703805 Chr.20: 56452192-56452192 on Build GRCh38
rs975696211 Chr.20: 56452249-56452249 on Build GRCh38
rs775018937 Chr.20: 56452330-56452330 on Build GRCh38
rs140450802 Chr.20: 56452152-56452152 on Build GRCh38
rs772751260 Chr.20: 56452159-56452159 on Build GRCh38
rs867712425 Chr.20: 56452170-56452170 on Build GRCh38
rs1048104148 Chr.20: 56452265-56452265 on Build GRCh38
rs756167828 Chr.20: 56452191-56452191 on Build GRCh38
rs140144032 Chr.20: 56452354-56452354 on Build GRCh38
rs76558241 Chr.20: 56452302-56452302 on Build GRCh38
rs779969415 Chr.20: 56452263-56452263 on Build GRCh38
rs767639107 Chr.20: 56452178-56452178 on Build GRCh38
rs142749187 Chr.20: 56452339-56452339 on Build GRCh38

View Full Product Details