Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 79784720-79784993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBX2
Entrez Gene ID
84733
Gene Name
chromobox 2
Gene Aliases
CDCA6, M33, SRXY5
UniGene
Hs.368410
Gene Chromosome Location
Chr.17: 79776254-79787650 on Build GRCh38
Transcripts
NM_005189.2 XM_011525382.1 XM_011525383.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2802777 c.1376G>A p.S459N Chr.17: 79784819-79784819 on Build GRCh38
5853698 c.1457C>T p.S486F Chr.17: 79784900-79784900 on Build GRCh38
2802779 c.1413C>T p.P471P Chr.17: 79784856-79784856 on Build GRCh38
2802782 c.1425G>A p.S475S Chr.17: 79784868-79784868 on Build GRCh38
309881 c.1441C>T p.Q481* Chr.17: 79784884-79784884 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148521974 Chr.17: 79784820-79784820 on Build GRCh38
rs782599629 Chr.17: 79784943-79784943 on Build GRCh38
rs1019545405 Chr.17: 79784955-79784955 on Build GRCh38
rs782445470 Chr.17: 79784862-79784862 on Build GRCh38
rs375182174 Chr.17: 79784952-79784952 on Build GRCh38
rs781932409 Chr.17: 79784852-79784852 on Build GRCh38
rs375416659 Chr.17: 79784881-79784881 on Build GRCh38
rs781856739 Chr.17: 79784856-79784856 on Build GRCh38
rs781942759 Chr.17: 79784895-79784895 on Build GRCh38
rs121908256 Chr.17: 79784771-79784771 on Build GRCh38
rs782386658 Chr.17: 79784847-79784847 on Build GRCh38
rs782464031 Chr.17: 79784941-79784941 on Build GRCh38
rs782635663 Chr.17: 79784865-79784865 on Build GRCh38
rs782234173 Chr.17: 79784843-79784843 on Build GRCh38
rs782450964 Chr.17: 79784745-79784745 on Build GRCh38
rs528502956 Chr.17: 79784901-79784901 on Build GRCh38
rs988082604 Chr.17: 79784828-79784828 on Build GRCh38
rs369849454 Chr.17: 79784891-79784891 on Build GRCh38
rs140909251 Chr.17: 79784871-79784871 on Build GRCh38
rs782652925 Chr.17: 79784959-79784959 on Build GRCh38
rs150658915 Chr.17: 79784864-79784864 on Build GRCh38
rs782167418 Chr.17: 79784824-79784824 on Build GRCh38
rs370780535 Chr.17: 79784748-79784748 on Build GRCh38
rs138828014 Chr.17: 79784781-79784781 on Build GRCh38
rs147956644 Chr.17: 79784944-79784944 on Build GRCh38
rs973656706 Chr.17: 79784801-79784801 on Build GRCh38
rs781924919 Chr.17: 79784766-79784766 on Build GRCh38
rs781789459 Chr.17: 79784932-79784932 on Build GRCh38
rs201172710 Chr.17: 79784770-79784770 on Build GRCh38
rs781926428 Chr.17: 79784965-79784965 on Build GRCh38
rs912043939 Chr.17: 79784844-79784844 on Build GRCh38
rs370084807 Chr.17: 79784812-79784812 on Build GRCh38
rs199566347 Chr.17: 79784799-79784799 on Build GRCh38
rs782717786 Chr.17: 79784863-79784863 on Build GRCh38
rs782014428 Chr.17: 79784850-79784850 on Build GRCh38
rs782334154 Chr.17: 79784833-79784833 on Build GRCh38
rs782732296 Chr.17: 79784791-79784791 on Build GRCh38
rs3751957 Chr.17: 79784892-79784892 on Build GRCh38
rs781911324 Chr.17: 79784859-79784859 on Build GRCh38
rs76915888 Chr.17: 79784798-79784798 on Build GRCh38
rs782670771 Chr.17: 79784741-79784741 on Build GRCh38
rs373385385 Chr.17: 79784902-79784902 on Build GRCh38
rs987728976 Chr.17: 79784950-79784950 on Build GRCh38
rs782287402 Chr.17: 79784756-79784756 on Build GRCh38
rs532091014 Chr.17: 79784757-79784757 on Build GRCh38
rs141949674 Chr.17: 79784787-79784787 on Build GRCh38
rs782275991 Chr.17: 79784960-79784960 on Build GRCh38
rs782235092 Chr.17: 79784946-79784946 on Build GRCh38
rs951060396 Chr.17: 79784949-79784949 on Build GRCh38
rs782818874 Chr.17: 79784910-79784910 on Build GRCh38
rs568793911 Chr.17: 79784792-79784792 on Build GRCh38
rs192165105 Chr.17: 79784835-79784835 on Build GRCh38
rs781978951 Chr.17: 79784774-79784774 on Build GRCh38
rs202081500 Chr.17: 79784868-79784868 on Build GRCh38
rs781786898 Chr.17: 79784743-79784743 on Build GRCh38
rs782242327 Chr.17: 79784872-79784872 on Build GRCh38
rs782096535 Chr.17: 79784896-79784896 on Build GRCh38
rs782567046 Chr.17: 79784870-79784870 on Build GRCh38
rs373878749 Chr.17: 79784782-79784782 on Build GRCh38
rs950986134 Chr.17: 79784827-79784827 on Build GRCh38
rs782268229 Chr.17: 79784867-79784867 on Build GRCh38
rs199672117 Chr.17: 79784857-79784857 on Build GRCh38
rs782088256 Chr.17: 79784768-79784768 on Build GRCh38
rs782422770 Chr.17: 79784962-79784962 on Build GRCh38
rs782349837 Chr.17: 79784973-79784973 on Build GRCh38
rs963046456 Chr.17: 79784913-79784913 on Build GRCh38
rs377749968 Chr.17: 79784969-79784969 on Build GRCh38
rs782657688 Chr.17: 79784755-79784755 on Build GRCh38

View Full Product Details