Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 42261192-42261417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EML4
Entrez Gene ID
27436
Gene Name
echinoderm microtubule associated protein like 4
Gene Aliases
C2orf2, ELP120, EMAP-4, EMAPL4, ROPP120
UniGene
Hs.713173
Gene Chromosome Location
Chr.2: 42169338-42332548 on Build GRCh38
Transcripts
NM_001145076.2 NM_019063.4 XM_005264267.1 XM_005264268.3 XM_006711991.1 XM_006711992.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
160695 c.425G>A p.R142Q Chr.2: 42261207-42261207 on Build GRCh38
5180262 c.512+7A>T p.? Chr.2: 42261301-42261301 on Build GRCh38
4920904 c.424C>T p.R142* Chr.2: 42261206-42261206 on Build GRCh38
4494480 c.433C>T p.P145S Chr.2: 42261215-42261215 on Build GRCh38
4680901 c.438_439insC p.Q148fs*32 Chr.2: 42261220-42261221 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368937633 Chr.2: 42261223-42261223 on Build GRCh38
rs757004510 Chr.2: 42261342-42261342 on Build GRCh38
rs771689075 Chr.2: 42261263-42261263 on Build GRCh38
rs896232700 Chr.2: 42261289-42261289 on Build GRCh38
rs372610199 Chr.2: 42261330-42261330 on Build GRCh38
rs775650829 Chr.2: 42261303-42261303 on Build GRCh38
rs777291102 Chr.2: 42261271-42261271 on Build GRCh38
rs534679626 Chr.2: 42261316-42261317 on Build GRCh38
rs141610396 Chr.2: 42261261-42261261 on Build GRCh38
rs745758418 Chr.2: 42261295-42261295 on Build GRCh38
rs183275581 Chr.2: 42261320-42261320 on Build GRCh38
rs1023256487 Chr.2: 42261316-42261316 on Build GRCh38
rs769871659 Chr.2: 42261301-42261301 on Build GRCh38
rs754435861 Chr.2: 42261327-42261327 on Build GRCh38
rs752852729 Chr.2: 42261215-42261215 on Build GRCh38
rs746613685 Chr.2: 42261339-42261339 on Build GRCh38
rs759190042 Chr.2: 42261315-42261319 on Build GRCh38
rs781082626 Chr.2: 42261345-42261345 on Build GRCh38
rs764301610 Chr.2: 42261306-42261306 on Build GRCh38
rs776611036 Chr.2: 42261288-42261288 on Build GRCh38
rs1017165903 Chr.2: 42261360-42261360 on Build GRCh38
rs201901757 Chr.2: 42261216-42261216 on Build GRCh38
rs529373671 Chr.2: 42261379-42261379 on Build GRCh38
rs774422635 Chr.2: 42261308-42261308 on Build GRCh38
rs778346266 Chr.2: 42261257-42261257 on Build GRCh38
rs756556695 Chr.2: 42261326-42261326 on Build GRCh38
rs767834432 Chr.2: 42261314-42261314 on Build GRCh38
rs143474845 Chr.2: 42261255-42261255 on Build GRCh38
rs549700611 Chr.2: 42261364-42261364 on Build GRCh38
rs565772763 Chr.2: 42261315-42261316 on Build GRCh38
rs746737127 Chr.2: 42261281-42261281 on Build GRCh38
rs962880886 Chr.2: 42261392-42261392 on Build GRCh38
rs762143576 Chr.2: 42261309-42261309 on Build GRCh38
rs763563612 Chr.2: 42261219-42261219 on Build GRCh38
rs377269900 Chr.2: 42261340-42261340 on Build GRCh38
rs563457625 Chr.2: 42261367-42261367 on Build GRCh38
rs1035911398 Chr.2: 42261334-42261335 on Build GRCh38
rs146741159 Chr.2: 42261304-42261304 on Build GRCh38
rs991634866 Chr.2: 42261391-42261391 on Build GRCh38
rs1052163835 Chr.2: 42261278-42261278 on Build GRCh38
rs781592633 Chr.2: 42261239-42261239 on Build GRCh38

View Full Product Details