Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 159196271-159196544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CADM3
Entrez Gene ID
57863
Gene Name
cell adhesion molecule 3
Gene Aliases
BIgR, IGSF4B, NECL1, Necl-1, TSLL1, synCAM3
UniGene
Hs.365689
Gene Chromosome Location
Chr.1: 159171587-159203142 on Build GRCh38
Transcripts
NM_001127173.1 NM_021189.3 NR_037870.1

Gene Symbol
CADM3-AS1
Entrez Gene ID
100131825
Gene Name
CADM3 antisense RNA 1
Gene Aliases
-
UniGene
Hs.701271
Gene Chromosome Location
Chr.1: 159195985-159202422 on Build GRCh38
Transcripts
NM_001127173.1 NM_021189.3 NR_037870.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1335326 c.824C>A p.P275H Chr.1: 159196394-159196394 on Build GRCh38
3976348 c.855G>C p.L285F Chr.1: 159196425-159196425 on Build GRCh38
1335327 c.873C>T p.R291R Chr.1: 159196443-159196443 on Build GRCh38
6618863 c.876C>T p.G292G Chr.1: 159196446-159196446 on Build GRCh38
897983 c.799C>T p.P267S Chr.1: 159196369-159196369 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774872813 Chr.1: 159196467-159196467 on Build GRCh38
rs760806925 Chr.1: 159196494-159196494 on Build GRCh38
rs138200155 Chr.1: 159196392-159196392 on Build GRCh38
rs767568858 Chr.1: 159196479-159196479 on Build GRCh38
rs1053818188 Chr.1: 159196438-159196438 on Build GRCh38
rs757871957 Chr.1: 159196396-159196396 on Build GRCh38
rs759993217 Chr.1: 159196476-159196476 on Build GRCh38
rs943244960 Chr.1: 159196507-159196507 on Build GRCh38
rs375293691 Chr.1: 159196329-159196329 on Build GRCh38
rs775563613 Chr.1: 159196484-159196484 on Build GRCh38
rs199882993 Chr.1: 159196322-159196322 on Build GRCh38
rs750427695 Chr.1: 159196495-159196495 on Build GRCh38
rs752876919 Chr.1: 159196340-159196340 on Build GRCh38
rs771249496 Chr.1: 159196465-159196465 on Build GRCh38
rs373766355 Chr.1: 159196443-159196443 on Build GRCh38
rs377679958 Chr.1: 159196460-159196460 on Build GRCh38
rs749658588 Chr.1: 159196453-159196453 on Build GRCh38
rs201711485 Chr.1: 159196360-159196360 on Build GRCh38
rs535252823 Chr.1: 159196429-159196429 on Build GRCh38
rs767429165 Chr.1: 159196400-159196400 on Build GRCh38
rs778876352 Chr.1: 159196376-159196376 on Build GRCh38
rs924611633 Chr.1: 159196395-159196395 on Build GRCh38
rs757070399 Chr.1: 159196363-159196363 on Build GRCh38
rs778864852 Chr.1: 159196470-159196477 on Build GRCh38
rs376129264 Chr.1: 159196413-159196413 on Build GRCh38
rs919592562 Chr.1: 159196302-159196302 on Build GRCh38
rs960219758 Chr.1: 159196321-159196321 on Build GRCh38
rs756337714 Chr.1: 159196345-159196345 on Build GRCh38
rs944216471 Chr.1: 159196365-159196365 on Build GRCh38
rs538364397 Chr.1: 159196338-159196338 on Build GRCh38
rs369002363 Chr.1: 159196334-159196334 on Build GRCh38
rs934754926 Chr.1: 159196431-159196431 on Build GRCh38
rs757194293 Chr.1: 159196467-159196468 on Build GRCh38
rs3026995 Chr.1: 159196518-159196518 on Build GRCh38
rs774384382 Chr.1: 159196377-159196377 on Build GRCh38
rs754096065 Chr.1: 159196506-159196506 on Build GRCh38
rs553949243 Chr.1: 159196323-159196323 on Build GRCh38
rs759640930 Chr.1: 159196336-159196336 on Build GRCh38
rs369549589 Chr.1: 159196442-159196442 on Build GRCh38
rs763823746 Chr.1: 159196346-159196346 on Build GRCh38

View Full Product Details