Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 68736857-68737110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCAR1
Entrez Gene ID
55749
Gene Name
cell division cycle and apoptosis regulator 1
Gene Aliases
-
UniGene
Hs.49853
Gene Chromosome Location
Chr.10: 68721144-68792377 on Build GRCh38
Transcripts
NM_001282959.1 NM_001282960.1 NM_018237.3 NR_104262.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2154310 c.246+1G>C p.? Chr.10: 68737049-68737049 on Build GRCh38
309887 c.74-1G>T p.? Chr.10: 68736875-68736875 on Build GRCh38
4626679 c.198T>C p.T66T Chr.10: 68737000-68737000 on Build GRCh38
4015309 c.210G>T p.Q70H Chr.10: 68737012-68737012 on Build GRCh38
74022 c.104T>C p.L35P Chr.10: 68736906-68736906 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566136047 Chr.10: 68736913-68736913 on Build GRCh38
rs182939954 Chr.10: 68736911-68736911 on Build GRCh38
rs772132235 Chr.10: 68736965-68736991 on Build GRCh38
rs770872831 Chr.10: 68736902-68736902 on Build GRCh38
rs761426078 Chr.10: 68737013-68737013 on Build GRCh38
rs774231215 Chr.10: 68736907-68736907 on Build GRCh38
rs760324023 Chr.10: 68737013-68737014 on Build GRCh38
rs998609110 Chr.10: 68737076-68737076 on Build GRCh38
rs1047452365 Chr.10: 68737073-68737073 on Build GRCh38
rs755253159 Chr.10: 68737035-68737035 on Build GRCh38
rs769989079 Chr.10: 68737078-68737078 on Build GRCh38
rs139946431 Chr.10: 68736950-68736950 on Build GRCh38
rs141356962 Chr.10: 68737028-68737028 on Build GRCh38
rs759525636 Chr.10: 68737010-68737010 on Build GRCh38
rs759329417 Chr.10: 68736949-68736949 on Build GRCh38
rs34601178 Chr.10: 68737022-68737022 on Build GRCh38
rs763632979 Chr.10: 68736998-68736998 on Build GRCh38
rs761772917 Chr.10: 68736886-68736886 on Build GRCh38
rs748286496 Chr.10: 68737009-68737009 on Build GRCh38
rs902788504 Chr.10: 68736990-68736990 on Build GRCh38
rs376968846 Chr.10: 68737021-68737021 on Build GRCh38
rs747302278 Chr.10: 68737006-68737006 on Build GRCh38
rs750906257 Chr.10: 68737004-68737004 on Build GRCh38
rs768967878 Chr.10: 68737008-68737008 on Build GRCh38
rs753816365 Chr.10: 68737067-68737069 on Build GRCh38
rs756682217 Chr.10: 68737003-68737003 on Build GRCh38
rs766348083 Chr.10: 68737022-68737030 on Build GRCh38
rs760417286 Chr.10: 68736988-68736988 on Build GRCh38
rs753411724 Chr.10: 68736999-68736999 on Build GRCh38
rs748259101 Chr.10: 68737072-68737072 on Build GRCh38
rs865874299 Chr.10: 68736917-68736918 on Build GRCh38
rs75794590 Chr.10: 68736892-68736892 on Build GRCh38
rs780634756 Chr.10: 68737005-68737005 on Build GRCh38
rs760340297 Chr.10: 68737012-68737012 on Build GRCh38
rs781481372 Chr.10: 68737044-68737044 on Build GRCh38
rs773493091 Chr.10: 68736982-68737016 on Build GRCh38
rs752474129 Chr.10: 68736959-68736959 on Build GRCh38
rs79020072 Chr.10: 68737016-68737016 on Build GRCh38

View Full Product Details