Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 174380565-174380830 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIR1
Entrez Gene ID
9541
Gene Name
corepressor interacting with RBPJ, 1
Gene Aliases
CIR
UniGene
Hs.632531
Gene Chromosome Location
Chr.2: 174348150-174395715 on Build GRCh38
Transcripts
NM_004882.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1009733 c.253-4T>G p.? Chr.2: 174380719-174380719 on Build GRCh38
7155425 c.265G>A p.E89K Chr.2: 174380703-174380703 on Build GRCh38
5394972 c.306C>T p.A102A Chr.2: 174380662-174380662 on Build GRCh38
3570106 c.252+2T>C p.? Chr.2: 174380802-174380802 on Build GRCh38
4582816 c.277G>A p.E93K Chr.2: 174380691-174380691 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766763122 Chr.2: 174380692-174380692 on Build GRCh38
rs200562118 Chr.2: 174380719-174380719 on Build GRCh38
rs952589595 Chr.2: 174380774-174380774 on Build GRCh38
rs753888108 Chr.2: 174380795-174380795 on Build GRCh38
rs748436436 Chr.2: 174380768-174380768 on Build GRCh38
rs149568348 Chr.2: 174380668-174380668 on Build GRCh38
rs753381459 Chr.2: 174380727-174380727 on Build GRCh38
rs373143920 Chr.2: 174380743-174380743 on Build GRCh38
rs767863653 Chr.2: 174380703-174380703 on Build GRCh38
rs565843308 Chr.2: 174380636-174380636 on Build GRCh38
rs749308506 Chr.2: 174380622-174380622 on Build GRCh38
rs748311598 Chr.2: 174380646-174380646 on Build GRCh38
rs749357380 Chr.2: 174380782-174380782 on Build GRCh38
rs918210157 Chr.2: 174380770-174380770 on Build GRCh38
rs370241477 Chr.2: 174380723-174380723 on Build GRCh38
rs755694876 Chr.2: 174380616-174380616 on Build GRCh38
rs759837671 Chr.2: 174380702-174380702 on Build GRCh38
rs754372739 Chr.2: 174380736-174380736 on Build GRCh38
rs1008961402 Chr.2: 174380670-174380670 on Build GRCh38
rs753467436 Chr.2: 174380700-174380702 on Build GRCh38
rs767103721 Chr.2: 174380612-174380612 on Build GRCh38
rs772628379 Chr.2: 174380794-174380794 on Build GRCh38
rs755821475 Chr.2: 174380722-174380722 on Build GRCh38
rs760016641 Chr.2: 174380793-174380793 on Build GRCh38
rs879123808 Chr.2: 174380625-174380625 on Build GRCh38
rs754698836 Chr.2: 174380610-174380610 on Build GRCh38
rs377577927 Chr.2: 174380697-174380697 on Build GRCh38
rs776863417 Chr.2: 174380643-174380643 on Build GRCh38
rs779758795 Chr.2: 174380755-174380755 on Build GRCh38
rs770990428 Chr.2: 174380786-174380786 on Build GRCh38
rs757262101 Chr.2: 174380729-174380729 on Build GRCh38
rs769867709 Chr.2: 174380658-174380658 on Build GRCh38
rs771861126 Chr.2: 174380640-174380640 on Build GRCh38
rs763563913 Chr.2: 174380691-174380691 on Build GRCh38
rs778749470 Chr.2: 174380738-174380738 on Build GRCh38
rs769980974 Chr.2: 174380771-174380771 on Build GRCh38
rs752345954 Chr.2: 174380615-174380615 on Build GRCh38
rs990889988 Chr.2: 174380764-174380764 on Build GRCh38
rs1014990050 Chr.2: 174380677-174380677 on Build GRCh38
rs757132114 Chr.2: 174380628-174380628 on Build GRCh38
rs758143266 Chr.2: 174380747-174380747 on Build GRCh38
rs961131578 Chr.2: 174380618-174380618 on Build GRCh38
rs376227677 Chr.2: 174380788-174380788 on Build GRCh38
rs534908530 Chr.2: 174380619-174380619 on Build GRCh38

View Full Product Details