Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 79778383-79778622 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBX2
Entrez Gene ID
84733
Gene Name
chromobox 2
Gene Aliases
CDCA6, M33, SRXY5
UniGene
Hs.368410
Gene Chromosome Location
Chr.17: 79776254-79787650 on Build GRCh38
Transcripts
NM_005189.2 NM_032647.3 XM_011525382.1 XM_011525383.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3388265 c.116+5G>A p.? Chr.17: 79778432-79778432 on Build GRCh38
3388264 c.116+5G>A p.? Chr.17: 79778432-79778432 on Build GRCh38
2802725 c.100C>T p.R34C Chr.17: 79778411-79778411 on Build GRCh38
4798746 c.75C>A p.G25G Chr.17: 79778386-79778386 on Build GRCh38
2802726 c.100C>T p.R34C Chr.17: 79778411-79778411 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs939674384 Chr.17: 79778514-79778514 on Build GRCh38
rs782313701 Chr.17: 79778434-79778434 on Build GRCh38
rs559189581 Chr.17: 79778542-79778542 on Build GRCh38
rs901385887 Chr.17: 79778531-79778531 on Build GRCh38
rs782791167 Chr.17: 79778458-79778458 on Build GRCh38
rs782688844 Chr.17: 79778433-79778434 on Build GRCh38
rs532823529 Chr.17: 79778585-79778585 on Build GRCh38
rs782353934 Chr.17: 79778437-79778437 on Build GRCh38
rs782673101 Chr.17: 79778422-79778422 on Build GRCh38
rs781983654 Chr.17: 79778451-79778451 on Build GRCh38
rs551178586 Chr.17: 79778597-79778597 on Build GRCh38
rs782073633 Chr.17: 79778435-79778435 on Build GRCh38
rs782296932 Chr.17: 79778423-79778423 on Build GRCh38
rs532462269 Chr.17: 79778582-79778582 on Build GRCh38
rs111795035 Chr.17: 79778596-79778596 on Build GRCh38
rs547345139 Chr.17: 79778496-79778496 on Build GRCh38
rs779295010 Chr.17: 79778558-79778558 on Build GRCh38
rs782622371 Chr.17: 79778411-79778411 on Build GRCh38
rs528874936 Chr.17: 79778476-79778476 on Build GRCh38
rs113008599 Chr.17: 79778483-79778483 on Build GRCh38
rs1041712628 Chr.17: 79778523-79778523 on Build GRCh38
rs782249643 Chr.17: 79778413-79778413 on Build GRCh38
rs781898210 Chr.17: 79778471-79778471 on Build GRCh38
rs997024288 Chr.17: 79778536-79778536 on Build GRCh38
rs782128610 Chr.17: 79778452-79778452 on Build GRCh38

View Full Product Details