Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73817390-73817661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMK2G
Entrez Gene ID
818
Gene Name
calcium/calmodulin dependent protein kinase II gamma
Gene Aliases
CAMK, CAMK-II, CAMKG
UniGene
Hs.523045
Gene Chromosome Location
Chr.10: 73812501-73874591 on Build GRCh38
Transcripts
NM_001204492.1 NM_001222.3 NM_001320898.1 NM_172169.2 NM_172170.4 NM_172171.2 NM_172173.2 XM_005270195.1 XM_005270197.1 XM_005270198.1 XM_005270199.1 XM_005270200.1 XM_005270201.1 XM_005270203.1 XM_005270205.1 XM_006717993.1 XM_006717996.1 XM_006717997.1 XM_006717998.1 XM_011540225.2 XM_017016716.1 XM_017016717.1 XM_017016718.1 XM_017016719.1 XM_017016720.1 XM_017016721.1 XM_017016722.1 XM_017016723.1 XM_017016724.1 XM_017016725.1 XM_017016726.1 XM_017016727.1 XM_017016728.1 XM_017016729.1 XM_017016730.1 XM_017016731.1 XM_017016732.1 XM_017016733.1 XM_017016734.1 XM_017016735.1 XM_017016736.1 XM_017016737.1 XM_017016738.1 XM_017016739.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1349079 c.1088G>A p.R363Q Chr.10: 73817551-73817551 on Build GRCh38
5050333 c.1305G>T p.L435L Chr.10: 73817517-73817517 on Build GRCh38
7105456 c.1281G>A p.E427E Chr.10: 73817541-73817541 on Build GRCh38
7105457 c.1194G>A p.E398E Chr.10: 73817541-73817541 on Build GRCh38
6619204 c.1343C>T p.T448M Chr.10: 73817479-73817479 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750350820 Chr.10: 73817572-73817572 on Build GRCh38
rs746361469 Chr.10: 73817637-73817637 on Build GRCh38
rs553576807 Chr.10: 73817590-73817590 on Build GRCh38
rs148234761 Chr.10: 73817611-73817611 on Build GRCh38
rs556044604 Chr.10: 73817415-73817415 on Build GRCh38
rs778753397 Chr.10: 73817563-73817563 on Build GRCh38
rs778942311 Chr.10: 73817446-73817446 on Build GRCh38
rs748388429 Chr.10: 73817463-73817463 on Build GRCh38
rs534533613 Chr.10: 73817455-73817456 on Build GRCh38
rs755069152 Chr.10: 73817574-73817574 on Build GRCh38
rs375277421 Chr.10: 73817509-73817509 on Build GRCh38
rs771265248 Chr.10: 73817599-73817599 on Build GRCh38
rs773571076 Chr.10: 73817476-73817476 on Build GRCh38
rs368242641 Chr.10: 73817456-73817456 on Build GRCh38
rs749695581 Chr.10: 73817595-73817595 on Build GRCh38
rs747087502 Chr.10: 73817452-73817452 on Build GRCh38
rs948980543 Chr.10: 73817517-73817517 on Build GRCh38
rs112344092 Chr.10: 73817454-73817455 on Build GRCh38
rs753624898 Chr.10: 73817431-73817431 on Build GRCh38
rs756806341 Chr.10: 73817450-73817451 on Build GRCh38
rs979232076 Chr.10: 73817414-73817414 on Build GRCh38
rs781326414 Chr.10: 73817584-73817584 on Build GRCh38
rs565730327 Chr.10: 73817455-73817455 on Build GRCh38
rs768747078 Chr.10: 73817453-73817453 on Build GRCh38
rs371949780 Chr.10: 73817484-73817484 on Build GRCh38
rs763255076 Chr.10: 73817479-73817479 on Build GRCh38
rs777932708 Chr.10: 73817591-73817591 on Build GRCh38
rs61864474 Chr.10: 73817615-73817615 on Build GRCh38
rs770210278 Chr.10: 73817467-73817467 on Build GRCh38
rs917546914 Chr.10: 73817508-73817508 on Build GRCh38
rs370530365 Chr.10: 73817538-73817538 on Build GRCh38
rs967908479 Chr.10: 73817558-73817558 on Build GRCh38
rs774891420 Chr.10: 73817490-73817490 on Build GRCh38
rs1020446415 Chr.10: 73817552-73817552 on Build GRCh38
rs763785684 Chr.10: 73817557-73817557 on Build GRCh38
rs757269896 Chr.10: 73817441-73817441 on Build GRCh38
rs890627766 Chr.10: 73817469-73817469 on Build GRCh38
rs757073848 Chr.10: 73817559-73817559 on Build GRCh38
rs201287315 Chr.10: 73817523-73817523 on Build GRCh38
rs760411599 Chr.10: 73817551-73817551 on Build GRCh38

View Full Product Details