Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 97024076-97024337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLX5
Entrez Gene ID
1749
Gene Name
distal-less homeobox 5
Gene Aliases
SHFM1D
UniGene
Hs.99348
Gene Chromosome Location
Chr.7: 97020390-97024831 on Build GRCh38
Transcripts
NM_005221.5 XM_005250185.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
287987 c.324C>T p.Y108Y Chr.7: 97024300-97024300 on Build GRCh38
6515276 c.296G>T p.S99I Chr.7: 97024328-97024328 on Build GRCh38
1698958 c.310T>A p.Y104N Chr.7: 97024314-97024314 on Build GRCh38
384688 c.311A>G p.Y104C Chr.7: 97024313-97024313 on Build GRCh38
6645136 c.296G>A p.S99N Chr.7: 97024328-97024328 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371127948 Chr.7: 97024301-97024301 on Build GRCh38
rs201376176 Chr.7: 97024291-97024291 on Build GRCh38
rs753621536 Chr.7: 97024296-97024296 on Build GRCh38
rs1015611109 Chr.7: 97024215-97024215 on Build GRCh38
rs921422739 Chr.7: 97024293-97024293 on Build GRCh38
rs200209992 Chr.7: 97024237-97024237 on Build GRCh38
rs985891661 Chr.7: 97024231-97024231 on Build GRCh38
rs1027464312 Chr.7: 97024200-97024200 on Build GRCh38
rs774697416 Chr.7: 97024279-97024279 on Build GRCh38
rs897283082 Chr.7: 97024155-97024155 on Build GRCh38
rs1013502068 Chr.7: 97024154-97024154 on Build GRCh38
rs781358596 Chr.7: 97024313-97024313 on Build GRCh38
rs756263693 Chr.7: 97024135-97024135 on Build GRCh38
rs535927739 Chr.7: 97024304-97024304 on Build GRCh38
rs530280507 Chr.7: 97024258-97024258 on Build GRCh38
rs767009962 Chr.7: 97024288-97024288 on Build GRCh38
rs768590287 Chr.7: 97024219-97024219 on Build GRCh38
rs971390524 Chr.7: 97024175-97024175 on Build GRCh38
rs780521417 Chr.7: 97024142-97024142 on Build GRCh38
rs201841970 Chr.7: 97024287-97024287 on Build GRCh38
rs778952255 Chr.7: 97024305-97024305 on Build GRCh38
rs79183163 Chr.7: 97024110-97024110 on Build GRCh38
rs200820594 Chr.7: 97024312-97024312 on Build GRCh38
rs773641315 Chr.7: 97024269-97024269 on Build GRCh38
rs766857960 Chr.7: 97024277-97024277 on Build GRCh38
rs1004637237 Chr.7: 97024184-97024184 on Build GRCh38
rs770291874 Chr.7: 97024257-97024257 on Build GRCh38
rs755697591 Chr.7: 97024292-97024292 on Build GRCh38
rs267601653 Chr.7: 97024311-97024311 on Build GRCh38
rs962752272 Chr.7: 97024259-97024259 on Build GRCh38
rs762108751 Chr.7: 97024240-97024240 on Build GRCh38
rs1025315222 Chr.7: 97024163-97024163 on Build GRCh38
rs763909664 Chr.7: 97024295-97024295 on Build GRCh38
rs763398794 Chr.7: 97024276-97024276 on Build GRCh38
rs750079834 Chr.7: 97024117-97024118 on Build GRCh38

View Full Product Details