Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 12664663-12664932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPPED1
Entrez Gene ID
55313
Gene Name
calcineurin like phosphoesterase domain containing 1
Gene Aliases
CSTP1
UniGene
Hs.460002
Gene Chromosome Location
Chr.16: 12659799-12803887 on Build GRCh38
Transcripts
NM_001099455.1 NM_018340.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1249046 c.909A>G p.I303M Chr.16: 12664922-12664922 on Build GRCh38
140085 c.916G>A p.D306N Chr.16: 12664915-12664915 on Build GRCh38
7400637 c.930G>C p.L310F Chr.16: 12664901-12664901 on Build GRCh38
4512945 c.921C>T p.L307L Chr.16: 12664910-12664910 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781239364 Chr.16: 12664865-12664865 on Build GRCh38
rs904417382 Chr.16: 12664840-12664840 on Build GRCh38
rs1046636697 Chr.16: 12664731-12664731 on Build GRCh38
rs755689329 Chr.16: 12664869-12664869 on Build GRCh38
rs367864945 Chr.16: 12664875-12664875 on Build GRCh38
rs537675910 Chr.16: 12664865-12664870 on Build GRCh38
rs755026639 Chr.16: 12664846-12664846 on Build GRCh38
rs562441710 Chr.16: 12664870-12664870 on Build GRCh38
rs143409742 Chr.16: 12664868-12664868 on Build GRCh38
rs751113154 Chr.16: 12664816-12664816 on Build GRCh38
rs773839952 Chr.16: 12664883-12664883 on Build GRCh38
rs754681484 Chr.16: 12664863-12664863 on Build GRCh38
rs1055254360 Chr.16: 12664714-12664714 on Build GRCh38
rs772298081 Chr.16: 12664885-12664885 on Build GRCh38
rs776101804 Chr.16: 12664895-12664895 on Build GRCh38
rs915728897 Chr.16: 12664716-12664716 on Build GRCh38
rs897410892 Chr.16: 12664742-12664742 on Build GRCh38
rs1042923594 Chr.16: 12664827-12664827 on Build GRCh38
rs753746918 Chr.16: 12664840-12664841 on Build GRCh38
rs948563540 Chr.16: 12664721-12664721 on Build GRCh38
rs746262783 Chr.16: 12664884-12664884 on Build GRCh38
rs994421943 Chr.16: 12664823-12664823 on Build GRCh38
rs887461503 Chr.16: 12664855-12664855 on Build GRCh38
rs781254681 Chr.16: 12664848-12664848 on Build GRCh38
rs369886431 Chr.16: 12664874-12664879 on Build GRCh38
rs765744731 Chr.16: 12664720-12664720 on Build GRCh38
rs373744185 Chr.16: 12664874-12664874 on Build GRCh38

View Full Product Details