Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 148859794-148860033 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPB1
Entrez Gene ID
1360
Gene Name
carboxypeptidase B1
Gene Aliases
CPB, PASP, PCPB
UniGene
Hs.477891
Gene Chromosome Location
Chr.3: 148827801-148860187 on Build GRCh38
Transcripts
NM_001871.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3002391 c.1073C>T p.A358V Chr.3: 148859821-148859821 on Build GRCh38
5039818 c.1082G>A p.G361D Chr.3: 148859830-148859830 on Build GRCh38
40815 c.1182C>T p.I394I Chr.3: 148859930-148859930 on Build GRCh38
4136974 c.1233C>T p.Y411Y Chr.3: 148859981-148859981 on Build GRCh38
3002394 c.1234G>A p.V412I Chr.3: 148859982-148859982 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs996451459 Chr.3: 148859899-148859899 on Build GRCh38
rs867981059 Chr.3: 148859869-148859869 on Build GRCh38
rs200398876 Chr.3: 148859955-148859955 on Build GRCh38
rs943478801 Chr.3: 148859933-148859933 on Build GRCh38
rs778069490 Chr.3: 148859839-148859839 on Build GRCh38
rs61733983 Chr.3: 148859930-148859930 on Build GRCh38
rs767732346 Chr.3: 148859837-148859837 on Build GRCh38
rs772650104 Chr.3: 148859894-148859894 on Build GRCh38
rs200456954 Chr.3: 148859838-148859838 on Build GRCh38
rs144356818 Chr.3: 148859887-148859887 on Build GRCh38
rs754761419 Chr.3: 148859825-148859825 on Build GRCh38
rs751289522 Chr.3: 148859820-148859820 on Build GRCh38
rs759354703 Chr.3: 148859920-148859920 on Build GRCh38
rs757430812 Chr.3: 148859950-148859950 on Build GRCh38
rs772369161 Chr.3: 148859886-148859886 on Build GRCh38
rs905645538 Chr.3: 148859823-148859823 on Build GRCh38
rs538633141 Chr.3: 148859942-148859942 on Build GRCh38
rs1054286417 Chr.3: 148859880-148859880 on Build GRCh38
rs963023009 Chr.3: 148859979-148859979 on Build GRCh38
rs758431117 Chr.3: 148859961-148859961 on Build GRCh38
rs1053337445 Chr.3: 148859970-148859970 on Build GRCh38
rs868343669 Chr.3: 148859905-148859905 on Build GRCh38
rs756873153 Chr.3: 148859931-148859931 on Build GRCh38
rs770815490 Chr.3: 148859900-148859900 on Build GRCh38
rs780338858 Chr.3: 148859968-148859968 on Build GRCh38
rs892083468 Chr.3: 148859981-148859981 on Build GRCh38
rs746191455 Chr.3: 148859867-148859867 on Build GRCh38
rs148833513 Chr.3: 148859943-148859943 on Build GRCh38
rs747820791 Chr.3: 148859891-148859891 on Build GRCh38
rs754387731 Chr.3: 148859840-148859840 on Build GRCh38
rs758782635 Chr.3: 148859960-148859960 on Build GRCh38
rs777434577 Chr.3: 148859997-148859997 on Build GRCh38
rs774294501 Chr.3: 148859904-148859904 on Build GRCh38
rs558275495 Chr.3: 148859846-148859846 on Build GRCh38
rs750930498 Chr.3: 148859959-148859959 on Build GRCh38
rs201519774 Chr.3: 148860001-148860001 on Build GRCh38
rs779589567 Chr.3: 148859864-148859864 on Build GRCh38
rs769279741 Chr.3: 148859893-148859893 on Build GRCh38
rs747412178 Chr.3: 148859982-148859982 on Build GRCh38
rs769369791 Chr.3: 148859993-148859993 on Build GRCh38
rs762454832 Chr.3: 148859896-148859896 on Build GRCh38
rs1042773404 Chr.3: 148860007-148860007 on Build GRCh38

View Full Product Details