Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 2898321-2898556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADD1
Entrez Gene ID
118
Gene Name
adducin 1
Gene Aliases
ADDA
UniGene
Hs.183706
Gene Chromosome Location
Chr.4: 2843732-2930076 on Build GRCh38
Transcripts
NM_001119.4 NM_001286645.1 NM_014189.3 NM_014190.3 NM_176801.2 XM_005247933.2 XM_005247934.2 XM_005247935.2 XM_005247936.2 XM_005247937.2 XM_017007703.1 XM_017007704.1 XM_017007705.1 XM_017007706.1 XM_017007707.1 XM_017007708.1 XM_017007709.1 XM_017007710.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1495796 c.899G>C p.R300P Chr.4: 2898446-2898446 on Build GRCh38
5047868 c.905A>G p.H302R Chr.4: 2898452-2898452 on Build GRCh38
5175811 c.899G>A p.R300Q Chr.4: 2898446-2898446 on Build GRCh38
3603275 c.960C>T p.N320N Chr.4: 2898507-2898507 on Build GRCh38
5990260 c.966G>A p.V322V Chr.4: 2898513-2898513 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780039355 Chr.4: 2898346-2898346 on Build GRCh38
rs566037365 Chr.4: 2898457-2898457 on Build GRCh38
rs780723149 Chr.4: 2898477-2898477 on Build GRCh38
rs750443352 Chr.4: 2898445-2898445 on Build GRCh38
rs765696659 Chr.4: 2898372-2898372 on Build GRCh38
rs1043552038 Chr.4: 2898407-2898407 on Build GRCh38
rs765316520 Chr.4: 2898427-2898427 on Build GRCh38
rs889184367 Chr.4: 2898413-2898413 on Build GRCh38
rs754031100 Chr.4: 2898420-2898420 on Build GRCh38
rs759997340 Chr.4: 2898387-2898387 on Build GRCh38
rs745783830 Chr.4: 2898399-2898399 on Build GRCh38
rs1006381446 Chr.4: 2898497-2898497 on Build GRCh38
rs747789048 Chr.4: 2898478-2898478 on Build GRCh38
rs551701080 Chr.4: 2898480-2898480 on Build GRCh38
rs868277399 Chr.4: 2898360-2898360 on Build GRCh38
rs764302827 Chr.4: 2898419-2898419 on Build GRCh38
rs147521356 Chr.4: 2898460-2898460 on Build GRCh38
rs754755610 Chr.4: 2898467-2898467 on Build GRCh38
rs746781628 Chr.4: 2898349-2898349 on Build GRCh38
rs768484562 Chr.4: 2898364-2898364 on Build GRCh38
rs772631725 Chr.4: 2898499-2898499 on Build GRCh38
rs761817740 Chr.4: 2898422-2898422 on Build GRCh38
rs747618884 Chr.4: 2898378-2898378 on Build GRCh38
rs374826670 Chr.4: 2898530-2898530 on Build GRCh38
rs926837565 Chr.4: 2898456-2898456 on Build GRCh38
rs761038171 Chr.4: 2898514-2898514 on Build GRCh38
rs774742702 Chr.4: 2898385-2898385 on Build GRCh38
rs776344429 Chr.4: 2898373-2898373 on Build GRCh38
rs372777117 Chr.4: 2898494-2898494 on Build GRCh38
rs1022451736 Chr.4: 2898347-2898347 on Build GRCh38
rs202124018 Chr.4: 2898446-2898446 on Build GRCh38
rs761021688 Chr.4: 2898415-2898415 on Build GRCh38
rs780129873 Chr.4: 2898459-2898459 on Build GRCh38
rs776755028 Chr.4: 2898475-2898475 on Build GRCh38
rs377154696 Chr.4: 2898389-2898389 on Build GRCh38
rs775890518 Chr.4: 2898404-2898404 on Build GRCh38
rs775906982 Chr.4: 2898501-2898501 on Build GRCh38

View Full Product Details