Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 18593654-18593887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTD1
Entrez Gene ID
92675
Gene Name
D-tyrosyl-tRNA deacylase 1
Gene Aliases
C20orf88, DUE-B, DUEB, HARS2, pqn-68
UniGene
Hs.659442
Gene Chromosome Location
Chr.20: 18587893-18763917 on Build GRCh38
Transcripts
NM_001318043.1 NM_080820.5 XM_005260872.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7424248 c.134T>A p.M45K Chr.20: 18593821-18593821 on Build GRCh38
3544438 c.47G>A p.G16E Chr.20: 18593734-18593734 on Build GRCh38
5500269 c.101C>T p.S34F Chr.20: 18593788-18593788 on Build GRCh38
5015657 c.70G>A p.G24R Chr.20: 18593757-18593757 on Build GRCh38
4141361 c.134+5G>T p.? Chr.20: 18593826-18593826 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753820181 Chr.20: 18593702-18593704 on Build GRCh38
rs113632385 Chr.20: 18593803-18593803 on Build GRCh38
rs35862400 Chr.20: 18593801-18593801 on Build GRCh38
rs760706395 Chr.20: 18593711-18593711 on Build GRCh38
rs766787842 Chr.20: 18593808-18593808 on Build GRCh38
rs755273398 Chr.20: 18593830-18593830 on Build GRCh38
rs907384808 Chr.20: 18593743-18593743 on Build GRCh38
rs754266231 Chr.20: 18593818-18593818 on Build GRCh38
rs777487984 Chr.20: 18593837-18593837 on Build GRCh38
rs536301437 Chr.20: 18593726-18593726 on Build GRCh38
rs758070863 Chr.20: 18593836-18593836 on Build GRCh38
rs746513796 Chr.20: 18593862-18593862 on Build GRCh38
rs779298681 Chr.20: 18593831-18593831 on Build GRCh38
rs146551342 Chr.20: 18593777-18593777 on Build GRCh38
rs771033535 Chr.20: 18593864-18593864 on Build GRCh38
rs756667078 Chr.20: 18593807-18593807 on Build GRCh38
rs1013184969 Chr.20: 18593685-18593685 on Build GRCh38
rs774457282 Chr.20: 18593784-18593784 on Build GRCh38
rs11545150 Chr.20: 18593754-18593754 on Build GRCh38
rs772783975 Chr.20: 18593710-18593710 on Build GRCh38
rs180699650 Chr.20: 18593722-18593722 on Build GRCh38
rs868148836 Chr.20: 18593800-18593800 on Build GRCh38
rs761982262 Chr.20: 18593791-18593791 on Build GRCh38
rs1049311464 Chr.20: 18593774-18593774 on Build GRCh38
rs1040394726 Chr.20: 18593828-18593828 on Build GRCh38
rs755088411 Chr.20: 18593861-18593866 on Build GRCh38
rs996241969 Chr.20: 18593846-18593846 on Build GRCh38
rs771742868 Chr.20: 18593693-18593693 on Build GRCh38
rs375190191 Chr.20: 18593833-18593833 on Build GRCh38
rs766892869 Chr.20: 18593840-18593840 on Build GRCh38

View Full Product Details