Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 34161028-34161267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABTB2
Entrez Gene ID
25841
Gene Name
ankyrin repeat and BTB domain containing 2
Gene Aliases
ABTB2A, BTBD22
UniGene
Hs.23361
Gene Chromosome Location
Chr.11: 34150987-34358008 on Build GRCh38
Transcripts
NM_145804.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3710171 c.2248G>A p.E750K Chr.11: 34161052-34161052 on Build GRCh38
7417223 c.2264C>T p.S755L Chr.11: 34161036-34161036 on Build GRCh38
3710172 c.1690G>A p.E564K Chr.11: 34161052-34161052 on Build GRCh38
7417224 c.1706C>T p.S569L Chr.11: 34161036-34161036 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755868794 Chr.11: 34161112-34161112 on Build GRCh38
rs767310161 Chr.11: 34161131-34161131 on Build GRCh38
rs907390204 Chr.11: 34161163-34161164 on Build GRCh38
rs913188336 Chr.11: 34161078-34161078 on Build GRCh38
rs747476865 Chr.11: 34161095-34161095 on Build GRCh38
rs771067701 Chr.11: 34161062-34161062 on Build GRCh38
rs887320265 Chr.11: 34161177-34161177 on Build GRCh38
rs772332320 Chr.11: 34161086-34161086 on Build GRCh38
rs201189199 Chr.11: 34161163-34161166 on Build GRCh38
rs768980552 Chr.11: 34161103-34161103 on Build GRCh38
rs528706679 Chr.11: 34161199-34161199 on Build GRCh38
rs922457392 Chr.11: 34161214-34161214 on Build GRCh38
rs772896450 Chr.11: 34161116-34161116 on Build GRCh38
rs975590360 Chr.11: 34161213-34161213 on Build GRCh38
rs751458530 Chr.11: 34161052-34161052 on Build GRCh38
rs532264500 Chr.11: 34161127-34161127 on Build GRCh38
rs1021980702 Chr.11: 34161181-34161181 on Build GRCh38
rs752712520 Chr.11: 34161133-34161133 on Build GRCh38
rs754817440 Chr.11: 34161128-34161128 on Build GRCh38
rs372289464 Chr.11: 34161235-34161235 on Build GRCh38
rs957609064 Chr.11: 34161172-34161172 on Build GRCh38
rs746060353 Chr.11: 34161076-34161076 on Build GRCh38
rs967204318 Chr.11: 34161114-34161114 on Build GRCh38
rs749270679 Chr.11: 34161059-34161059 on Build GRCh38
rs931345942 Chr.11: 34161167-34161167 on Build GRCh38
rs761186172 Chr.11: 34161112-34161112 on Build GRCh38
rs368385988 Chr.11: 34161159-34161159 on Build GRCh38
rs369634146 Chr.11: 34161113-34161113 on Build GRCh38
rs868632955 Chr.11: 34161173-34161173 on Build GRCh38
rs375860359 Chr.11: 34161124-34161124 on Build GRCh38
rs1041580851 Chr.11: 34161200-34161200 on Build GRCh38
rs919912638 Chr.11: 34161160-34161160 on Build GRCh38
rs762680569 Chr.11: 34161118-34161118 on Build GRCh38
rs900017148 Chr.11: 34161203-34161203 on Build GRCh38
rs777214599 Chr.11: 34161106-34161106 on Build GRCh38
rs768953354 Chr.11: 34161169-34161169 on Build GRCh38
rs552766755 Chr.11: 34161134-34161134 on Build GRCh38
rs775823176 Chr.11: 34161087-34161087 on Build GRCh38
rs750175782 Chr.11: 34161110-34161111 on Build GRCh38
rs377511923 Chr.11: 34161053-34161053 on Build GRCh38
rs963774997 Chr.11: 34161190-34161190 on Build GRCh38

View Full Product Details