Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62705390-62705651 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSCL2
Entrez Gene ID
26580
Gene Name
BSCL2, seipin lipid droplet biogenesis associated
Gene Aliases
GNG3LG, HMN5, PELD, SPG17
UniGene
Hs.533709
Gene Chromosome Location
Chr.11: 62690262-62709619 on Build GRCh38
Transcripts
NM_001122955.3 NM_001130702.2 NM_032667.6 NR_037948.1 NR_037949.1 NR_037946.1

Gene Symbol
HNRNPUL2-BSCL2
Entrez Gene ID
100534595
Gene Name
HNRNPUL2-BSCL2 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.533709
Gene Chromosome Location
Chr.11: 62690262-62727384 on Build GRCh38
Transcripts
NM_001122955.3 NM_001130702.2 NM_032667.6 NR_037948.1 NR_037949.1 NR_037946.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6134017 c.57C>T p.V19V Chr.11: 62705456-62705456 on Build GRCh38
4796647 c.196G>A p.V66I Chr.11: 62705509-62705509 on Build GRCh38
6040482 c.67C>T p.R23C Chr.11: 62705446-62705446 on Build GRCh38
5964982 c.137G>C p.R46T Chr.11: 62705568-62705568 on Build GRCh38
4756381 c.115C>A p.Q39K Chr.11: 62705590-62705590 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781626368 Chr.11: 62705429-62705429 on Build GRCh38
rs756668260 Chr.11: 62705435-62705435 on Build GRCh38
rs572867847 Chr.11: 62705432-62705432 on Build GRCh38
rs3763853 Chr.11: 62705572-62705572 on Build GRCh38
rs907887190 Chr.11: 62705505-62705505 on Build GRCh38
rs759201886 Chr.11: 62705481-62705481 on Build GRCh38
rs776497908 Chr.11: 62705478-62705478 on Build GRCh38
rs777422952 Chr.11: 62705548-62705548 on Build GRCh38
rs953445189 Chr.11: 62705423-62705423 on Build GRCh38
rs867357580 Chr.11: 62705475-62705475 on Build GRCh38
rs894136605 Chr.11: 62705598-62705598 on Build GRCh38
rs948421905 Chr.11: 62705607-62705607 on Build GRCh38
rs775351747 Chr.11: 62705580-62705580 on Build GRCh38
rs769536524 Chr.11: 62705574-62705574 on Build GRCh38
rs771754989 Chr.11: 62705440-62705440 on Build GRCh38
rs201493373 Chr.11: 62705581-62705581 on Build GRCh38
rs774441432 Chr.11: 62705591-62705591 on Build GRCh38
rs772832629 Chr.11: 62705448-62705448 on Build GRCh38
rs767605984 Chr.11: 62705605-62705605 on Build GRCh38
rs140762669 Chr.11: 62705500-62705500 on Build GRCh38
rs756907468 Chr.11: 62705521-62705521 on Build GRCh38
rs761845256 Chr.11: 62705596-62705596 on Build GRCh38
rs749865861 Chr.11: 62705535-62705535 on Build GRCh38
rs770641122 Chr.11: 62705458-62705458 on Build GRCh38
rs755534269 Chr.11: 62705541-62705541 on Build GRCh38
rs756478297 Chr.11: 62705517-62705517 on Build GRCh38
rs746722132 Chr.11: 62705456-62705456 on Build GRCh38
rs775340076 Chr.11: 62705485-62705485 on Build GRCh38
rs11558433 Chr.11: 62705484-62705484 on Build GRCh38
rs879253928 Chr.11: 62705433-62705433 on Build GRCh38
rs767589914 Chr.11: 62705512-62705512 on Build GRCh38
rs531137749 Chr.11: 62705589-62705589 on Build GRCh38
rs757487761 Chr.11: 62705501-62705501 on Build GRCh38
rs750662693 Chr.11: 62705629-62705629 on Build GRCh38
rs900971126 Chr.11: 62705575-62705575 on Build GRCh38
rs746461003 Chr.11: 62705555-62705555 on Build GRCh38
rs780387759 Chr.11: 62705439-62705439 on Build GRCh38
rs764226928 Chr.11: 62705496-62705496 on Build GRCh38
rs770638595 Chr.11: 62705557-62705557 on Build GRCh38
rs542073179 Chr.11: 62705568-62705568 on Build GRCh38
rs762865520 Chr.11: 62705486-62705486 on Build GRCh38
rs750833941 Chr.11: 62705516-62705516 on Build GRCh38
rs568354548 Chr.11: 62705420-62705420 on Build GRCh38

View Full Product Details