Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 127657683-127657920 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTXN3
Entrez Gene ID
613212
Gene Name
cortexin 3
Gene Aliases
KABE
UniGene
Hs.66194
Gene Chromosome Location
Chr.5: 127649021-127658630 on Build GRCh38
Transcripts
NM_001048252.2 NM_001127385.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1060267 c.178C>T p.P60S Chr.5: 127657699-127657699 on Build GRCh38
378791 c.236C>A p.A79D Chr.5: 127657757-127657757 on Build GRCh38
3135794 c.169C>T p.R57* Chr.5: 127657690-127657690 on Build GRCh38
3608145 c.170G>A p.R57Q Chr.5: 127657691-127657691 on Build GRCh38
256709 c.229G>A p.D77N Chr.5: 127657750-127657750 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766889034 Chr.5: 127657742-127657742 on Build GRCh38
rs148234170 Chr.5: 127657841-127657841 on Build GRCh38
rs750001642 Chr.5: 127657743-127657743 on Build GRCh38
rs745983302 Chr.5: 127657797-127657797 on Build GRCh38
rs763274176 Chr.5: 127657804-127657804 on Build GRCh38
rs147610610 Chr.5: 127657732-127657732 on Build GRCh38
rs781109503 Chr.5: 127657793-127657793 on Build GRCh38
rs779750938 Chr.5: 127657755-127657755 on Build GRCh38
rs771072325 Chr.5: 127657824-127657824 on Build GRCh38
rs777970815 Chr.5: 127657744-127657744 on Build GRCh38
rs368464050 Chr.5: 127657704-127657704 on Build GRCh38
rs570359779 Chr.5: 127657706-127657706 on Build GRCh38
rs962230108 Chr.5: 127657813-127657813 on Build GRCh38
rs761841777 Chr.5: 127657741-127657741 on Build GRCh38
rs114243151 Chr.5: 127657825-127657825 on Build GRCh38
rs775798287 Chr.5: 127657799-127657799 on Build GRCh38
rs370866029 Chr.5: 127657722-127657722 on Build GRCh38
rs780172652 Chr.5: 127657826-127657826 on Build GRCh38
rs908276778 Chr.5: 127657840-127657840 on Build GRCh38
rs775391633 Chr.5: 127657729-127657729 on Build GRCh38
rs781683976 Chr.5: 127657776-127657777 on Build GRCh38
rs986068366 Chr.5: 127657855-127657855 on Build GRCh38
rs753050412 Chr.5: 127657758-127657758 on Build GRCh38
rs762905924 Chr.5: 127657730-127657730 on Build GRCh38
rs367856467 Chr.5: 127657770-127657770 on Build GRCh38
rs747430809 Chr.5: 127657786-127657786 on Build GRCh38
rs536490791 Chr.5: 127657772-127657772 on Build GRCh38
rs545518034 Chr.5: 127657864-127657864 on Build GRCh38
rs142310306 Chr.5: 127657749-127657749 on Build GRCh38
rs778860466 Chr.5: 127657782-127657782 on Build GRCh38
rs771488710 Chr.5: 127657791-127657791 on Build GRCh38

View Full Product Details