Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 86440099-86440368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCA2
Entrez Gene ID
9635
Gene Name
chloride channel accessory 2
Gene Aliases
CACC, CACC3, CLCRG2, CaCC-3
UniGene
Hs.241551
Gene Chromosome Location
Chr.1: 86424086-86456558 on Build GRCh38
Transcripts
NM_006536.5 XM_011542448.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
682846 c.1357C>A p.L453M Chr.1: 86440301-86440301 on Build GRCh38
3493047 c.1340C>T p.S447L Chr.1: 86440284-86440284 on Build GRCh38
6629771 c.1224A>G p.G408G Chr.1: 86440168-86440168 on Build GRCh38
5524749 c.1282A>G p.N428D Chr.1: 86440226-86440226 on Build GRCh38
4401145 c.1352C>T p.P451L Chr.1: 86440296-86440296 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572565439 Chr.1: 86440316-86440316 on Build GRCh38
rs745342946 Chr.1: 86440291-86440298 on Build GRCh38
rs202177831 Chr.1: 86440174-86440174 on Build GRCh38
rs757965959 Chr.1: 86440150-86440150 on Build GRCh38
rs537020575 Chr.1: 86440193-86440193 on Build GRCh38
rs746250353 Chr.1: 86440176-86440176 on Build GRCh38
rs898155479 Chr.1: 86440146-86440146 on Build GRCh38
rs752106592 Chr.1: 86440246-86440246 on Build GRCh38
rs780824760 Chr.1: 86440267-86440267 on Build GRCh38
rs960443104 Chr.1: 86440194-86440194 on Build GRCh38
rs776652671 Chr.1: 86440324-86440324 on Build GRCh38
rs781738328 Chr.1: 86440175-86440175 on Build GRCh38
rs764351218 Chr.1: 86440335-86440335 on Build GRCh38
rs974721364 Chr.1: 86440292-86440292 on Build GRCh38
rs202107894 Chr.1: 86440331-86440331 on Build GRCh38
rs759635583 Chr.1: 86440325-86440325 on Build GRCh38
rs764594002 Chr.1: 86440243-86440243 on Build GRCh38
rs979275803 Chr.1: 86440285-86440285 on Build GRCh38
rs775305371 Chr.1: 86440339-86440339 on Build GRCh38
rs145061827 Chr.1: 86440305-86440305 on Build GRCh38
rs768334665 Chr.1: 86440298-86440298 on Build GRCh38
rs755747239 Chr.1: 86440280-86440280 on Build GRCh38
rs1022166469 Chr.1: 86440284-86440284 on Build GRCh38
rs757836190 Chr.1: 86440173-86440173 on Build GRCh38
rs191164171 Chr.1: 86440224-86440224 on Build GRCh38
rs757895186 Chr.1: 86440258-86440258 on Build GRCh38
rs752250550 Chr.1: 86440141-86440141 on Build GRCh38
rs771163204 Chr.1: 86440208-86440208 on Build GRCh38
rs763300782 Chr.1: 86440334-86440334 on Build GRCh38
rs768834782 Chr.1: 86440200-86440200 on Build GRCh38
rs749048932 Chr.1: 86440290-86440290 on Build GRCh38
rs764964319 Chr.1: 86440138-86440138 on Build GRCh38
rs763240287 Chr.1: 86440239-86440239 on Build GRCh38
rs766314705 Chr.1: 86440211-86440211 on Build GRCh38
rs141340884 Chr.1: 86440287-86440287 on Build GRCh38
rs771809953 Chr.1: 86440293-86440293 on Build GRCh38
rs749990472 Chr.1: 86440271-86440271 on Build GRCh38
rs777125786 Chr.1: 86440160-86440160 on Build GRCh38
rs376243564 Chr.1: 86440204-86440204 on Build GRCh38
rs753056318 Chr.1: 86440236-86440236 on Build GRCh38
rs150391383 Chr.1: 86440317-86440317 on Build GRCh38
rs112347137 Chr.1: 86440245-86440245 on Build GRCh38
rs138147089 Chr.1: 86440326-86440326 on Build GRCh38
rs149017440 Chr.1: 86440221-86440221 on Build GRCh38
rs748547263 Chr.1: 86440188-86440188 on Build GRCh38
rs199964844 Chr.1: 86440216-86440216 on Build GRCh38
rs986704815 Chr.1: 86440321-86440321 on Build GRCh38
rs747578568 Chr.1: 86440162-86440162 on Build GRCh38
rs1023184191 Chr.1: 86440203-86440203 on Build GRCh38
rs770238442 Chr.1: 86440183-86440183 on Build GRCh38
rs145870955 Chr.1: 86440205-86440205 on Build GRCh38
rs148776402 Chr.1: 86440232-86440232 on Build GRCh38

View Full Product Details