Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 50048518-50048755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEFB112
Entrez Gene ID
245915
Gene Name
defensin beta 112
Gene Aliases
DEFB-12
UniGene
Hs.571092
Gene Chromosome Location
Chr.6: 50043575-50048651 on Build GRCh38
Transcripts
NM_001037498.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3628361 c.3G>A p.M1I Chr.6: 50048649-50048649 on Build GRCh38
6106889 c.108C>A p.I36I Chr.6: 50048544-50048544 on Build GRCh38
6106890 c.113C>A p.T38K Chr.6: 50048539-50048539 on Build GRCh38
4893002 c.106A>T p.I36F Chr.6: 50048546-50048546 on Build GRCh38
1697371 c.100G>A p.E34K Chr.6: 50048552-50048552 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs963416410 Chr.6: 50048720-50048720 on Build GRCh38
rs773977541 Chr.6: 50048669-50048669 on Build GRCh38
rs190024828 Chr.6: 50048586-50048586 on Build GRCh38
rs766735842 Chr.6: 50048703-50048703 on Build GRCh38
rs777992885 Chr.6: 50048661-50048661 on Build GRCh38
rs769099493 Chr.6: 50048655-50048655 on Build GRCh38
rs780624440 Chr.6: 50048590-50048590 on Build GRCh38
rs377262797 Chr.6: 50048677-50048677 on Build GRCh38
rs759773356 Chr.6: 50048658-50048659 on Build GRCh38
rs761872417 Chr.6: 50048687-50048687 on Build GRCh38
rs769328567 Chr.6: 50048603-50048603 on Build GRCh38
rs371555913 Chr.6: 50048634-50048634 on Build GRCh38
rs779333966 Chr.6: 50048561-50048562 on Build GRCh38
rs111458001 Chr.6: 50048681-50048681 on Build GRCh38
rs760438909 Chr.6: 50048631-50048631 on Build GRCh38
rs1043337458 Chr.6: 50048615-50048615 on Build GRCh38
rs560045743 Chr.6: 50048564-50048564 on Build GRCh38
rs752525417 Chr.6: 50048630-50048630 on Build GRCh38
rs773888964 Chr.6: 50048607-50048607 on Build GRCh38
rs767149002 Chr.6: 50048624-50048624 on Build GRCh38
rs1055232267 Chr.6: 50048589-50048589 on Build GRCh38
rs780395533 Chr.6: 50048650-50048650 on Build GRCh38
rs539374567 Chr.6: 50048697-50048701 on Build GRCh38
rs778997201 Chr.6: 50048636-50048636 on Build GRCh38
rs866401740 Chr.6: 50048557-50048557 on Build GRCh38
rs147546237 Chr.6: 50048653-50048653 on Build GRCh38
rs925250603 Chr.6: 50048644-50048644 on Build GRCh38
rs373797808 Chr.6: 50048600-50048600 on Build GRCh38
rs781764854 Chr.6: 50048656-50048656 on Build GRCh38
rs756107012 Chr.6: 50048560-50048560 on Build GRCh38
rs73739915 Chr.6: 50048671-50048671 on Build GRCh38
rs200405124 Chr.6: 50048632-50048632 on Build GRCh38
rs753733566 Chr.6: 50048683-50048683 on Build GRCh38
rs750560210 Chr.6: 50048689-50048689 on Build GRCh38
rs771759638 Chr.6: 50048674-50048674 on Build GRCh38
rs777654118 Chr.6: 50048563-50048563 on Build GRCh38
rs927741020 Chr.6: 50048561-50048561 on Build GRCh38
rs144576723 Chr.6: 50048551-50048551 on Build GRCh38
rs773081528 Chr.6: 50048676-50048676 on Build GRCh38
rs758717051 Chr.6: 50048640-50048640 on Build GRCh38
rs770407491 Chr.6: 50048605-50048605 on Build GRCh38
rs368572348 Chr.6: 50048700-50048700 on Build GRCh38
rs779039259 Chr.6: 50048566-50048566 on Build GRCh38
rs748627626 Chr.6: 50048666-50048666 on Build GRCh38
rs762400186 Chr.6: 50048604-50048604 on Build GRCh38
rs757293157 Chr.6: 50048635-50048635 on Build GRCh38
rs375047649 Chr.6: 50048638-50048638 on Build GRCh38
rs759287245 Chr.6: 50048608-50048608 on Build GRCh38

View Full Product Details