Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 58150858-58151099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C20orf85
Entrez Gene ID
128602
Gene Name
chromosome 20 open reading frame 85
Gene Aliases
LLC1, bA196N14.1
UniGene
Hs.43977
Gene Chromosome Location
Chr.20: 58150927-58161130 on Build GRCh38
Transcripts
NM_178456.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1198609 c.5C>T p.A2V Chr.20: 58150969-58150969 on Build GRCh38
3673143 c.48T>G p.L16L Chr.20: 58151012-58151012 on Build GRCh38
1727345 c.68+1G>T p.? Chr.20: 58151033-58151033 on Build GRCh38
1412696 c.21C>T p.S7S Chr.20: 58150985-58150985 on Build GRCh38
5794379 c.42G>A p.M14I Chr.20: 58151006-58151006 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs998637043 Chr.20: 58151075-58151075 on Build GRCh38
rs375410038 Chr.20: 58151037-58151037 on Build GRCh38
rs367679911 Chr.20: 58150911-58150911 on Build GRCh38
rs755182831 Chr.20: 58150927-58150927 on Build GRCh38
rs763524747 Chr.20: 58151050-58151051 on Build GRCh38
rs763833144 Chr.20: 58150960-58150960 on Build GRCh38
rs907822000 Chr.20: 58151010-58151010 on Build GRCh38
rs780060007 Chr.20: 58151056-58151056 on Build GRCh38
rs372144544 Chr.20: 58150913-58150913 on Build GRCh38
rs561812689 Chr.20: 58150938-58150938 on Build GRCh38
rs746769200 Chr.20: 58151063-58151063 on Build GRCh38
rs964109827 Chr.20: 58150939-58150939 on Build GRCh38
rs1011787413 Chr.20: 58151079-58151079 on Build GRCh38
rs367946142 Chr.20: 58150925-58150925 on Build GRCh38
rs771924570 Chr.20: 58150945-58150945 on Build GRCh38
rs949129344 Chr.20: 58150984-58150984 on Build GRCh38
rs567427257 Chr.20: 58151074-58151074 on Build GRCh38
rs775914009 Chr.20: 58150953-58150953 on Build GRCh38
rs111861700 Chr.20: 58150916-58150916 on Build GRCh38
rs369862453 Chr.20: 58151047-58151047 on Build GRCh38
rs975445368 Chr.20: 58150961-58150961 on Build GRCh38
rs201080740 Chr.20: 58151053-58151053 on Build GRCh38
rs113991855 Chr.20: 58150896-58150896 on Build GRCh38
rs759709413 Chr.20: 58151055-58151055 on Build GRCh38
rs202223558 Chr.20: 58151054-58151054 on Build GRCh38
rs562607871 Chr.20: 58151002-58151002 on Build GRCh38
rs1017187636 Chr.20: 58150924-58150924 on Build GRCh38
rs906967821 Chr.20: 58150880-58150880 on Build GRCh38
rs1020809649 Chr.20: 58150885-58150885 on Build GRCh38
rs776044937 Chr.20: 58150979-58150979 on Build GRCh38
rs761338769 Chr.20: 58150988-58150988 on Build GRCh38
rs1001274182 Chr.20: 58150893-58150893 on Build GRCh38
rs775293343 Chr.20: 58150947-58150947 on Build GRCh38
rs957504525 Chr.20: 58150910-58150910 on Build GRCh38
rs755236306 Chr.20: 58151004-58151004 on Build GRCh38
rs761235156 Chr.20: 58151080-58151080 on Build GRCh38
rs765695598 Chr.20: 58150989-58150989 on Build GRCh38
rs973295052 Chr.20: 58150940-58150940 on Build GRCh38
rs1034532411 Chr.20: 58150894-58150894 on Build GRCh38
rs770184907 Chr.20: 58150952-58150953 on Build GRCh38
rs193186455 Chr.20: 58150940-58150940 on Build GRCh38
rs764658728 Chr.20: 58151053-58151054 on Build GRCh38
rs774292690 Chr.20: 58150941-58150941 on Build GRCh38
rs200557259 Chr.20: 58151024-58151024 on Build GRCh38
rs766789436 Chr.20: 58151001-58151001 on Build GRCh38
rs940598441 Chr.20: 58151029-58151029 on Build GRCh38
rs778718322 Chr.20: 58151031-58151031 on Build GRCh38
rs540321417 Chr.20: 58150929-58150929 on Build GRCh38
rs901239463 Chr.20: 58150908-58150908 on Build GRCh38
rs200156567 Chr.20: 58151038-58151038 on Build GRCh38
rs750876827 Chr.20: 58150990-58150990 on Build GRCh38
rs781458885 Chr.20: 58151009-58151009 on Build GRCh38
rs544405573 Chr.20: 58150959-58150959 on Build GRCh38
rs780260446 Chr.20: 58150918-58150918 on Build GRCh38
rs6070358 Chr.20: 58151051-58151051 on Build GRCh38
rs921770174 Chr.20: 58150977-58150977 on Build GRCh38
rs369261647 Chr.20: 58150944-58150944 on Build GRCh38
rs386815621 Chr.20: 58151051-58151054 on Build GRCh38
rs768457703 Chr.20: 58151068-58151068 on Build GRCh38
rs557763159 Chr.20: 58150915-58150915 on Build GRCh38
rs34488967 Chr.20: 58150968-58150969 on Build GRCh38
rs758777362 Chr.20: 58150997-58150997 on Build GRCh38
rs748195417 Chr.20: 58151015-58151015 on Build GRCh38

View Full Product Details