Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 149741532-149741767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMMD2
Entrez Gene ID
51122
Gene Name
COMM domain containing 2
Gene Aliases
HSPC042
UniGene
Hs.432729
Gene Chromosome Location
Chr.3: 149737160-149752507 on Build GRCh38
Transcripts
NM_016094.3 NR_135149.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7423843 c.458A>T p.H153L Chr.3: 149741663-149741663 on Build GRCh38
1693583 c.490G>A p.V164I Chr.3: 149741631-149741631 on Build GRCh38
5653982 c.495G>A p.L165L Chr.3: 149741626-149741626 on Build GRCh38
7223281 c.567T>C p.N189N Chr.3: 149741554-149741554 on Build GRCh38
5496482 c.403-5T>C p.? Chr.3: 149741723-149741723 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200137592 Chr.3: 149741630-149741630 on Build GRCh38
rs769037268 Chr.3: 149741586-149741586 on Build GRCh38
rs953299938 Chr.3: 149741647-149741647 on Build GRCh38
rs760434689 Chr.3: 149741620-149741620 on Build GRCh38
rs143514325 Chr.3: 149741619-149741619 on Build GRCh38
rs963419043 Chr.3: 149741632-149741632 on Build GRCh38
rs778530127 Chr.3: 149741678-149741678 on Build GRCh38
rs748900756 Chr.3: 149741655-149741655 on Build GRCh38
rs148182261 Chr.3: 149741679-149741679 on Build GRCh38
rs752087488 Chr.3: 149741636-149741636 on Build GRCh38
rs780747363 Chr.3: 149741635-149741635 on Build GRCh38
rs7633238 Chr.3: 149741554-149741554 on Build GRCh38
rs375560161 Chr.3: 149741731-149741731 on Build GRCh38
rs758547862 Chr.3: 149741634-149741634 on Build GRCh38
rs763900101 Chr.3: 149741625-149741625 on Build GRCh38
rs765422645 Chr.3: 149741628-149741628 on Build GRCh38
rs999365636 Chr.3: 149741605-149741605 on Build GRCh38
rs369806421 Chr.3: 149741732-149741732 on Build GRCh38
rs781583579 Chr.3: 149741640-149741640 on Build GRCh38
rs945967300 Chr.3: 149741689-149741689 on Build GRCh38
rs11549572 Chr.3: 149741626-149741626 on Build GRCh38
rs772060164 Chr.3: 149741611-149741611 on Build GRCh38
rs755324646 Chr.3: 149741638-149741638 on Build GRCh38
rs768649706 Chr.3: 149741608-149741608 on Build GRCh38
rs748917874 Chr.3: 149741728-149741732 on Build GRCh38
rs199801530 Chr.3: 149741680-149741680 on Build GRCh38
rs373916335 Chr.3: 149741595-149741595 on Build GRCh38
rs954785183 Chr.3: 149741696-149741696 on Build GRCh38
rs750591347 Chr.3: 149741633-149741633 on Build GRCh38
rs780696013 Chr.3: 149741559-149741559 on Build GRCh38
rs139970043 Chr.3: 149741602-149741602 on Build GRCh38
rs747400364 Chr.3: 149741580-149741580 on Build GRCh38
rs746918136 Chr.3: 149741600-149741600 on Build GRCh38
rs1546732 Chr.3: 149741590-149741590 on Build GRCh38
rs770625334 Chr.3: 149741673-149741673 on Build GRCh38

View Full Product Details